Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abnova™ Human PRR13 Full-length ORF (ADZ15792.1, 1 a.a. - 148 a.a.) Recombinant Protein with GST-tag at N-terminal

Used for AP, Array, ELISA, WB-Re

Specifications

Name Human PRR13 Full-length ORF (ADZ15792.1, 1 a.a. - 148 a.a.) Recombinant Protein with GST-tag at N-terminal
Recombinant Yes
Purification Method Glutathione Sepharose 4 Fast Flow
For Use With (Application) AP, Array, ELISA, WB-Re
Molecular Weight 42.7kDa
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
89-958-377
View Documents
Abnova™
H00054458P01L
25μg This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000. N/A
89-958-376
View Documents
Abnova™
H00054458P01S
10μg This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000. N/A
Description

Description

Sequence: MWNPNAGQPGPNPYPPNIGCPGGSNPAHPPPINPPFPPGPCPPPPGAPHGNPAFPPGGPPHPVPQPGYPGCQPLGPYPPPYPPPAPGIPPVNPLAPGMVGPAVIVDKKMQKKMKKAHKKMHKHQKHHKYHKHGKHSSSSSSSSSSDSD
Specifications

Specifications

Human PRR13 Full-length ORF (ADZ15792.1, 1 a.a. - 148 a.a.) Recombinant Protein with GST-tag at N-terminal
Glutathione Sepharose 4 Fast Flow
42.7kDa
Yes
AP, Array, ELISA, WB-Re
Store at −80°C. Aliquot to avoid repeated freezing and thawing.
SDS
Product Certifications
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit