Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abnova™ Human PRR4 Partial ORF (NP_009175.1, 17 a.a. - 116 a.a.) Recombinant Protein with GST-tag at N-terminal

Used for AP, Array, ELISA, WB-Re

$460.50 - $709.50

Specifications

Name PRR4 (Human) Recombinant Protein (Q01)
Recombinant Yes
For Use With (Application) Antibody Production, Enzyme-linked Immunoabsorbent Assay, Protein Array, Western Blot (Recombinant protein)
Molecular Weight 36.74kDa
Storage Requirements Store at -80°C. Aliquot to avoid repeated freezing and thawing.
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
89-958-378
View Documents
Abnova™
H00011272Q01S
10 ug
Each for $460.50
Add to cart
 
89-958-379
View Documents
Abnova™
H00011272Q01L
25 ug
Each for $709.50
Add to cart
 
Description

Description

Lacrimal proline rich protein is a member of the proline-rich protein family which lacks a conserved repetitive domain. It may have a role in protective functions in the eye. Two alternatively spliced transcript variants that encode different proteins have been described for this gene. [provided by RefSeq]

Sequence: QSTDNDVNYEDFTFTIPDVEDSSQRPDQGPQRPPPEGLLPRPPGDSGNQDDGPQQRPPKPGGHHRHPPPPPFQNQQRPPQRGHRQLSLPRFPSVSLQEAS
Specifications

Specifications

PRR4 (Human) Recombinant Protein (Q01)
Antibody Production, Enzyme-linked Immunoabsorbent Assay, Protein Array, Western Blot (Recombinant protein)
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
Yes
36.74kDa
SDS
Product Certifications
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit