Learn More
Abnova™ Human PSCD4 Full-length ORF (NP_037517.1, 1 a.a. - 394 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
$499.00 - $769.00
Specifications
Accession Number | NP_037517.1 |
---|---|
For Use With (Application) | Antibody Production, Protein Array, ELISA, Western Blot |
Formulation | 50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. |
Gene ID (Entrez) | 27128 |
Molecular Weight (g/mol) | 72.1kDa |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
89-958-475
|
Abnova™
H00027128P01L |
25 ug |
Each for $769.00
|
|
|||||
89-958-474
|
Abnova™
H00027128P01S |
10 ug |
Each for $499.00
|
|
|||||
Description
The protein encoded by this gene is a member of the PSCD family. Members of this family have identical structural organization that consists of an N-terminal coiled-coil motif, a central Sec7 domain, and a C-terminal pleckstrin homology (PH) domain. The coiled-coil motif is involved in homodimerization, the Sec7 domain contains guanine-nucleotide exchange protein (GEP) activity, and the PH domain interacts with phospholipids and is responsible for association of PSCDs with membranes. Members of this family appear to mediate the regulation of protein sorting and membrane trafficking. The encoded protein exhibits GEP activity in vitro with both ARF1 and ARF5 but is inactive with ARF6. The structures of this gene and CYTH1 are very similar. [provided by RefSeq]
Sequence: MDLCHPEPAELSSGETEELQRIKWHRKQLLEDIQKLKDEIADVFAQIDCFESAEESRMAQKEKELCIGRKKFNMDPAKGIQYFIEHKLLTPDVQDIARFLYKGEGLNKTAIGTYLGERDPINLQVLQAFVDCHEFANLNLVQALRQFLWSFRLPGEAQKIDRMMEAFATRYCLCNPGVFQSTDTCYVLSFSIIMLNTSLHNPNVRDRPPFERFVSMNRGINNGSDLPEDQLRNLFDSIKSEPFSIPEDDGNDLTHTFFNPDREGWLLKLGGRVKTWKRRWFILTDNCLYYFEFTTDKEPRGIIPLENLSVQKVDDPKKPFCLELYNPSCRGQKIKACKTDGDGRVVEGKHESYRISATSAEERDQWIESIRASITRVPFYDLVSTRKKKIASKQSpecifications
NP_037517.1 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
72.1kDa | |
Glutathione Sepharose 4 Fast Flow | |
MDLCHPEPAELSSGETEELQRIKWHRKQLLEDIQKLKDEIADVFAQIDCFESAEESRMAQKEKELCIGRKKFNMDPAKGIQYFIEHKLLTPDVQDIARFLYKGEGLNKTAIGTYLGERDPINLQVLQAFVDCHEFANLNLVQALRQFLWSFRLPGEAQKIDRMMEAFATRYCLCNPGVFQSTDTCYVLSFSIIMLNTSLHNPNVRDRPPFERFVSMNRGINNGSDLPEDQLRNLFDSIKSEPFSIPEDDGNDLTHTFFNPDREGWLLKLGGRVKTWKRRWFILTDNCLYYFEFTTDKEPRGIIPLENLSVQKVDDPKKPFCLELYNPSCRGQKIKACKTDGDGRVVEGKHESYRISATSAEERDQWIESIRASITRVPFYDLVSTRKKKIASKQ | |
RUO | |
CYTH4 | |
Wheat Germ (in vitro) | |
GST | |
Liquid |
Antibody Production, Protein Array, ELISA, Western Blot | |
27128 | |
PSCD4 (Human) Recombinant Protein (P01) | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
CYT4/DJ63G5.1/PSCD4 | |
CYTH4 | |
Recombinant | |
wheat germ expression system |
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.