Learn More
Novus Biologicals™ Human PSGL-1/CD162 ELISA Kit
ELISA kits are ideal for the quantification of target antigens in tissue extracts, serum and cell culture. 
Manufacturer:  Novus Biologicals™ NBP191256
Catalog No. NBP191256
Add to cart
Description
Description
The Human PSGL-1/CD162 ELISA Kit (Colorimetric) measures human PSGL-1/CD162 in cell culture supernatant, serum.
Specifications
Specifications
1.6 - 50 U/mL
6404
96 Tests
Cell culture supernatant, serum
Storage of components varies. See protocol for specific instructions.
CD Markers, Cholesterol Metabolism, Immunology, Lipid and Metabolism
CD162, CD162 antigen, P-selectin glycoprotein ligand 1, PSGL1, PSGL-1cutaneous lymphocyte-associated associated antigen, selectin P ligandCLA
ELISA
ELISA Kit
1.0 U/mL
Human (Hu)
PSGL-1/CD162
SELPLG