Learn More
Abnova Human PSMF1 (NP_006805, 1 a.a. - 271 a.a.) Full-length Recombinant Protein with His-tag
Used for Func, SDS-PAGE
Manufacturer: Abnova P4540
Description
The 26S proteasome is a multicatalytic proteinase complex with a highly ordered structure composed of 2 complexes, a 20S core and a 19S regulator. The 20S core is composed of 4 rings of 28 non-identical subunits; 2 rings are composed of 7 alpha subunits and 2 rings are composed of 7 beta subunits. The 19S regulator is composed of a base, which contains 6 ATPase subunits and 2 non-ATPase subunits, and a lid, which contains up to 10 non-ATPase subunits. Proteasomes are distributed throughout eukaryotic cells at a high concentration and cleave peptides in an ATP/ubiquitin-dependent process in a non-lysosomal pathway. An essential function of a modified proteasome, the immunoproteasome, is the processing of class I MHC peptides. This gene encodes a protein that inhibits the activation of the proteasome by the 11S and 19S regulators. Alternative transcript variants have been identified for this gene. [provided by RefSeq]
Sequence: MGSSHHHHHHSSGLVPRGSHMAGLEVLFASAAPAITCRQDALVCFLHWEVVTHGYCGLGVGDQPGPNDKKSELLPAGWNNNKDLYVLRYEYKDGSRKLLVKAITVESSMILNVLEYGSQQVADLTLNLDDYIDAEHLGDFHRTYKNSEELRSRIVSGIITPIHEQWEKANVSSPHREFPPATAREVDPLRIPPHHPHTSRQPPWCDPLGPFVVGGEDLDPFGPRRGGMIVDPLRSGFPRALIDPSSGLPNRLPPGAVPPGARFDPFGPIGTSPPGPNPDHLPPPGYDDMYLSpecifications
NP_006805 | |
0.5 mg/mL | |
Liquid | |
9491 | |
PSMF1 (Human) Recombinant Protein | |
>85% by SDS - PAGE | |
100 ug | |
RUO | |
PSMF1 | |
Escherichia coli | |
His |
Escherichia coli expression system | |
Functional Study, Sodium Dodecyl Sulfate–Polyacrylamide Gel Electrophoresis | |
Liquid | |
31.9kDa | |
Escherichia coli expression system | |
15% SDS-PAGE Stained with Coomassie Blue | |
Store at -20°C. For long term storage store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
PI31 | |
PSMF1 | |
Yes |
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.