Learn More
Abnova Human PTPDC1 Full-length ORF (AAH67120.1, 1 a.a. - 754 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
Manufacturer: Abnova H00138639P01L
Description
The protein encoded by this gene contains a characteristic motif of protein tyrosine phosphatases (PTPs). PTPs regulate activities of phosphoproteins through dephosphorylation. They are signaling molecules involved in the regulation of a wide variety of biological processes. The specific function of this protein has not yet been determined. Alternatively spliced transcript variants encoding distinct isoforms have been identified. [provided by RefSeq]
Sequence: MAAGVLPQNEQPYSTLVNNSECVANMKGNLERPTPKYTKVGERLRHVIPGHMACSMACGGRACKYENPARWSEQEQAIKGVYSSWVTDNILAMARPSSELLEKYHIIDQFLSHGIKTIINLQRPGEHASCGNPLEQESGFTYLPEAFMEAGIYFYNFGWKDYGVASLTTILDMVKVMTFALQEGKVAIHCHAGLGRTGVLIACYLVFATRMTADQAIIFVRAKRPNSIQTRGQLLCVREFTQFLTPLRNIFSCCDPKAHAVTLPQYLIRQRHLLHGYEARLLKHVPKIIHLVCKLLLDLAENRPVMMKDVSEGPGLSAEIEKTMSEMVTMQLDKELLRHDSDFSNPPNPTAVAADFDNRGMIFSNEQQFDPLWKRRNVECLQPLTHLKRRLSYSDSDLKRAENLLEQGETPQTVPAQILVGHKPRQQKLISHCYIPQSPEPDLHKEALVRSTLSFWSQSKFGGLEGLKDNGSPIFHGRIIPKEAQQSGAFSADVSGSHSPGEPVSPSFANVHKDPNPAHQQVSHCQCKTHGVGSPGSVRQNSRTPRSPLDCGSSPKAQFLVEHETQDSKDLSEAASHSALQSELSAEARRILAAKALANLNESVEKEELKRKVEMWQKELNSRDGAWERICGERDPFILCSLMWSWVEQLKEPVITKEDVDMLVDRRADAAEALFLLEKGQHQTILCVLHCIVNLQTIPVDVEEAFLAHAIKAFTKVNFDSENGPTVYNTLKKIFKHTLEEKRKMTKDGPKPGLSpecifications
AAH67120.1 | |
Antibody Production, Array, Enzyme-linked Immunoabsorbent Assay, Western Blot (Recombinant protein) | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
110.9kDa | |
Glutathione Sepharose 4 Fast Flow | |
25 ug | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
FLJ42922|PTP9Q22 | |
PTPDC1 | |
Yes |
wheat germ expression system | |
Liquid | |
138639 | |
PTPDC1 (Human) Recombinant Protein (P01) | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
MAAGVLPQNEQPYSTLVNNSECVANMKGNLERPTPKYTKVGERLRHVIPGHMACSMACGGRACKYENPARWSEQEQAIKGVYSSWVTDNILAMARPSSELLEKYHIIDQFLSHGIKTIINLQRPGEHASCGNPLEQESGFTYLPEAFMEAGIYFYNFGWKDYGVASLTTILDMVKVMTFALQEGKVAIHCHAGLGRTGVLIACYLVFATRMTADQAIIFVRAKRPNSIQTRGQLLCVREFTQFLTPLRNIFSCCDPKAHAVTLPQYLIRQRHLLHGYEARLLKHVPKIIHLVCKLLLDLAENRPVMMKDVSEGPGLSAEIEKTMSEMVTMQLDKELLRHDSDFSNPPNPTAVAADFDNRGMIFSNEQQFDPLWKRRNVECLQPLTHLKRRLSYSDSDLKRAENLLEQGETPQTVPAQILVGHKPRQQKLISHCYIPQSPEPDLHKEALVRSTLSFWSQSKFGGLEGLKDNGSPIFHGRIIPKEAQQSGAFSADVSGSHSPGEPVSPSFANVHKDPNPAHQQVSHCQCKTHGVGSPGSVRQNSRTPRSPLDCGSSPKAQFLVEHETQDSKDLSEAASHSALQSELSAEARRILAAKALANLNESVEKEELKRKVEMWQKELNSRDGAWERICGERDPFILCSLMWSWVEQLKEPVITKEDVDMLVDRRADAAEALFLLEKGQHQTILCVLHCIVNLQTIPVDVEEAFLAHAIKAFTKVNFDSENGPTVYNTLKKIFKHTLEEKRKMTKDGPKPGL | |
RUO | |
PTPDC1 | |
Wheat Germ (in vitro) | |
GST |
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.