Learn More
Abnova Human PTPN4 Partial ORF (NP_002821, 361 a.a. - 460 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
Manufacturer: Abnova H00005775Q01L
Description
The protein encoded by this gene is a member of the protein tyrosine phosphatase (PTP) family. PTPs are known to be signaling molecules that regulate a variety of cellular processes including cell growth, differentiation, mitotic cycle, and oncogenic transformation. This protein contains a C-terminal PTP domain and an N-terminal domain homologous to the band 4.1 superfamily of cytoskeletal-associated proteins. This PTP has been shown to interact with glutamate receptor delta 2 and epsilon subunits, and is thought to play a role in signalling downstream of the glutamate receptors through tyrosine dephosphorylation. [provided by RefSeq]
Sequence: KPLARKLMDWEVVSRNSISDDRLETQSLPSRSPPGTPNHRNSTFTQEGTRLRPSSVGHLVDHMVHTSPSEVFVNQRSPSSTQANSIVLESSPSQETPGDGSpecifications
NP_002821 | |
Antibody Production, Enzyme-linked Immunoabsorbent Assay, Protein Array, Western Blot (Recombinant protein) | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
36.74kDa | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
KPLARKLMDWEVVSRNSISDDRLETQSLPSRSPPGTPNHRNSTFTQEGTRLRPSSVGHLVDHMVHTSPSEVFVNQRSPSSTQANSIVLESSPSQETPGDG | |
RUO | |
PTPN4 | |
Wheat Germ (in vitro) | |
GST |
wheat germ expression system | |
Liquid | |
5775 | |
PTPN4 (Human) Recombinant Protein (Q01) | |
25 ug | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
PTPMEG|PTPMEG1 | |
PTPN4 | |
Yes |
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.