Learn More
Abnova Human PTPRO Full-length ORF (AAH35960, 1 a.a. - 597 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
Manufacturer: Abnova H00005800P01L
Description
This gene encodes a receptor-type protein tyrosine phosphatase containing a single intracellular catalytic domain with a characteristic signature motif. The gene product, which has a transmembrane domain, is an integral membrane protein. Several alternatively spliced transcript variants, some of which encode different isoforms of the protein, have been described. These variants exhibit tissue-specific expression. [provided by RefSeq]
Sequence: MGHLPTGIHGARRLLPLLWLFVLFKNATAFHVTVQDDNNIVVSLEASDVISPASVYVVKITGESKNYFFEFEEFNSTLPPPVIFKASYHGLYYIITLVVVNGNVVTKPSRSITVLTKPLPVTSVSIYDYKPSPETGVLFEIHYPEKYNVFTRVNISYWEGKDFRTMLYKDFFKGKTVFNHWLPGMCYSNITFQLVSEATFNKSTLVEYSGVSHEPKQHRTAPYPPQNISVRIVNLNKNNWEEQSGNFPEESFMRSQDTIGKEKLFHFTEETPEIPSGNISSGWPDFNSSDYETTSQPYWWDSASAAPESEDEFVSVLPMEYENNSTLSETEKSTSGSFSFFPVQMILTWLPPKPPTAFDGFHIHIEREENFTEYLMVDEEAHEFVAELKEPGKYKLSVTTFSSSGSCETRKSQSAKSLSFYISPSGEWIEELTEKPQHVSVHVLSSTTALMSWTSSQENYNSTIVSVVSLTCQKQKESQRLEKQYCTQVNSSKPIIENLVPGAQYQVVIYLRKGPLIGPPSDPVTFAIVPTGIKDLMLYPLGPTAVVLSWTRPYLGVFRKYVVEMFYFNPATMTSEWTTYYEIAATVSLTASVVIFPSpecifications
AAH35960 | |
Antibody Production, Array, Enzyme-linked Immunoabsorbent Assay, Western Blot (Recombinant protein) | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
91.41kDa | |
Glutathione Sepharose 4 Fast Flow | |
25 ug | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
GLEPP1|PTP-U2|PTPU2 | |
PTPRO | |
Yes |
wheat germ expression system | |
Liquid | |
5800 | |
PTPRO (Human) Recombinant Protein (P01) | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
MGHLPTGIHGARRLLPLLWLFVLFKNATAFHVTVQDDNNIVVSLEASDVISPASVYVVKITGESKNYFFEFEEFNSTLPPPVIFKASYHGLYYIITLVVVNGNVVTKPSRSITVLTKPLPVTSVSIYDYKPSPETGVLFEIHYPEKYNVFTRVNISYWEGKDFRTMLYKDFFKGKTVFNHWLPGMCYSNITFQLVSEATFNKSTLVEYSGVSHEPKQHRTAPYPPQNISVRIVNLNKNNWEEQSGNFPEESFMRSQDTIGKEKLFHFTEETPEIPSGNISSGWPDFNSSDYETTSQPYWWDSASAAPESEDEFVSVLPMEYENNSTLSETEKSTSGSFSFFPVQMILTWLPPKPPTAFDGFHIHIEREENFTEYLMVDEEAHEFVAELKEPGKYKLSVTTFSSSGSCETRKSQSAKSLSFYISPSGEWIEELTEKPQHVSVHVLSSTTALMSWTSSQENYNSTIVSVVSLTCQKQKESQRLEKQYCTQVNSSKPIIENLVPGAQYQVVIYLRKGPLIGPPSDPVTFAIVPTGIKDLMLYPLGPTAVVLSWTRPYLGVFRKYVVEMFYFNPATMTSEWTTYYEIAATVSLTASVVIFP | |
RUO | |
PTPRO | |
Wheat Germ (in vitro) | |
GST |
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.