Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Novus Biologicals™ Human Pyruvate Dehydrogenase E1-alpha subunit ELISA Kit (Colorimetric)

This kit recognizes Human Pyruvate Dehydrogenase E1-alpha subunit in samples.

$629.00

Specifications

For Use With (Application) ELISA
Gene ID (Entrez) 5160
Host Species Human
Product Type ELISA Kit (Colorimetric)
Quantity 1 Kit
View More Specs

Products 2
Catalog Number Mfr. No. Assay Range Assay Sensitivity Price Quantity & Availability  
Catalog Number Mfr. No. Assay Range Assay Sensitivity Price Quantity & Availability  
NB016779
View Documents
Novus Biologicals™
NBP281220
0.16 to 10 ng/mL 0.09 ng/mL
Each for $629.00
Add to cart
 
NB016777
View Documents
Novus Biologicals™
NBP281218
31.25 to 2000 pg/mL 18.75 pg/mL
Each for $629.00
Add to cart
 
Description

Description

No significant cross-reactivity or interference between Human Pyruvate Dehydrogenase E1-alpha subunit and analogues was observed.
Specifications

Specifications

ELISA
Human
1 Kit
Serum, Plasma, Cell Supernatant, Other Biological Fluids
Pyruvate Dehydrogenase E1-alpha subunit
5160
ELISA Kit (Colorimetric)
Cancer, Core ESC Like Genes, Lipid and Metabolism, Phospho Specific, Signal Transduction, Stem Cell Markers
Storage is content dependent
SDS
Product Certifications
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit