Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human Pyruvate Dehydrogenase Phosphatase/PDP1 Antibody, R&D Systems™

Sheep Polyclonal Antibody
Supplier: R&D Systems AF7260SP
This item is not returnable.
View return policy
Description
Pyruvate Dehydrogenase Phosphatase/PDP1/PPM2C Polyclonal specifically detects Pyruvate Dehydrogenase Phosphatase/PDP1/PPM2C in Human samples. It is validated for Western Blot.Specifications
Pyruvate Dehydrogenase Phosphatase/PDP1/PPM2C | |
Polyclonal | |
Unconjugated | |
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
[Pyruvate dehydrogenase [acetyl-transferring]]-phosphatase 1, mitochondrial, EC 3.1.3, EC 3.1.3.43, MGC119646, PDH, PDP 1, PDP1, PDPC, PDPC 1, PDPFLJ32517, PPM2C, PPM2CFLJ56179, Protein phosphatase 2C, protein phosphatase 2C, magnesium-dependent, catalytic subunit, pyruvate dehydrogenase (Lipoamide) phosphatase-phosphatase, Pyruvate dehydrogenase phosphatase catalytic subunit 1, pyruvate dehyrogenase phosphatase catalytic subunit 1 | |
Sheep | |
Affinity Purified | |
RUO | |
54704 | |
Sterile PBS to a final concentration of 0.2 mg/mL. | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
Western Blot | |
LYOPH | |
Western Blot 1 ug/mL | |
Q9P0J1 | |
PDP1 | |
E. coli-derived recombinant human Pyruvate Dehydrogenase Phosphatase/PDP1 Ala72-Glu537 Accession # Q9P0J1 | |
25 μg | |
Primary | |
Detects human Pyruvate Dehydrogenase Phosphatase/PDP1 in direct ELISAs and Western blots. | |
Human | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction