Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Abnova Human RAB11FIP1 Full-length ORF (NP_001002233.1, 1 a.a. - 612 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
Manufacturer: Abnova H00080223P01L
Description
Proteins of the large Rab GTPase family (see RAB1A; MIM 179508) have regulatory roles in the formation, targeting, and fusion of intracellular transport vesicles. RAB11FIP1 is one of many proteins that interact with and regulate Rab GTPases (Hales et al., 2001 [PubMed 11495908]).[supplied by OMIM]
Sequence: MGRTRETGTEKNTSSLELEESLPEQPETGRQEEELPRFPCKKQDYSPSSGEAQEVPFALSLSSDGAVSPVGELAAGGDRDLESQAGSLVESKARDAAEEVAPPLPMGASVPSIDSMMRKLEEMGLNLRKDQKKTKKRVSFSEQLFTEEAVAGAALLVEGHSSCPQELNPAWSVAGNASDGEPPESPHAEDSERESVTTPGPATCGAPASPADHLLLPSQEESFSEVPMSEASSAKDTPLFRMEGEDALVTQYQSKASDHEGLLSDPLSDLQLVSDFKSPIMADLNLSLPSIPEVASDDERIDQVEDDGDQVEDDGETAKSSTLDIGALSLGLVVPCPERGKGPSGEADRLVLGEGLCDFRLQAPQASVTAPSEQTTEFGIHKPHLGKSSSLDKQLPGPSGGEEEKPMGNGSPSPPPGTSLDNPVPSPSPSEIFPVTHSFPSSAHSDTHHTSTAESQKKATAEGSAGRVENFGKRKPLLQAWVSPSETHPVSAQPGAGTGSAKHRLHPVKPMNAMATKVANCSLGTATIISENLNNEVMMKKYSPSDPAFAYAQLTHDELIQLVLKQKETISKKEFQVRELEDYIDNLLVRVMEETPNILRIPTQVGKKAGKMSpecifications
NP_001002233.1 | |
Antibody Production, Array, Enzyme-linked Immunoabsorbent Assay, Western Blot (Recombinant protein) | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
91.6kDa | |
Glutathione Sepharose 4 Fast Flow | |
25 ug | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
DKFZp686E2214|FLJ22524|FLJ22622|MGC78448|NOEL1A|RCP|rab11-FIP1 | |
RAB11FIP1 | |
Yes |
wheat germ expression system | |
Liquid | |
80223 | |
RAB11FIP1 (Human) Recombinant Protein (P01) | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
MGRTRETGTEKNTSSLELEESLPEQPETGRQEEELPRFPCKKQDYSPSSGEAQEVPFALSLSSDGAVSPVGELAAGGDRDLESQAGSLVESKARDAAEEVAPPLPMGASVPSIDSMMRKLEEMGLNLRKDQKKTKKRVSFSEQLFTEEAVAGAALLVEGHSSCPQELNPAWSVAGNASDGEPPESPHAEDSERESVTTPGPATCGAPASPADHLLLPSQEESFSEVPMSEASSAKDTPLFRMEGEDALVTQYQSKASDHEGLLSDPLSDLQLVSDFKSPIMADLNLSLPSIPEVASDDERIDQVEDDGDQVEDDGETAKSSTLDIGALSLGLVVPCPERGKGPSGEADRLVLGEGLCDFRLQAPQASVTAPSEQTTEFGIHKPHLGKSSSLDKQLPGPSGGEEEKPMGNGSPSPPPGTSLDNPVPSPSPSEIFPVTHSFPSSAHSDTHHTSTAESQKKATAEGSAGRVENFGKRKPLLQAWVSPSETHPVSAQPGAGTGSAKHRLHPVKPMNAMATKVANCSLGTATIISENLNNEVMMKKYSPSDPAFAYAQLTHDELIQLVLKQKETISKKEFQVRELEDYIDNLLVRVMEETPNILRIPTQVGKKAGKM | |
RUO | |
RAB11FIP1 | |
Wheat Germ (in vitro) | |
GST |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title