Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Abnova™ Human RAB4B Full-length ORF (NP_057238.3, 1 a.a. - 213 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
Supplier: Abnova™ H00053916P01L
Description
RAB proteins, such as RAB4B, are members of the RAS superfamily of small GTPases that are involved in vesicular trafficking (He et al., 2002 [PubMed 12450215]).[supplied by OMIM]
Sequence: MAETYDFLFKFLVIGSAGTGKSCLLHQFIENKFKQDSNHTIGVEFGSRVVNVGGKTVKLQIWDTAGQERFRSVTRSYYRGAAGALLVYDITSRETYNSLAAWLTDARTLASPNIVVILCGNKKDLDPEREVTFLEASRFAQENELMFLETSALTGENVEEAFLKCARTILNKIDSGELDPERMGSGIQYGDASLRQLRQPRSAQAVAPQPCGCSpecifications
NP_057238.3 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
50kDa | |
Glutathione Sepharose 4 Fast Flow | |
25 ug | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
FLJ78649/MGC52123 | |
RAB4B | |
Recombinant | |
wheat germ expression system |
Antibody Production, Protein Array, ELISA, Western Blot | |
53916 | |
RAB4B (Human) Recombinant Protein (P01) | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
MAETYDFLFKFLVIGSAGTGKSCLLHQFIENKFKQDSNHTIGVEFGSRVVNVGGKTVKLQIWDTAGQERFRSVTRSYYRGAAGALLVYDITSRETYNSLAAWLTDARTLASPNIVVILCGNKKDLDPEREVTFLEASRFAQENELMFLETSALTGENVEEAFLKCARTILNKIDSGELDPERMGSGIQYGDASLRQLRQPRSAQAVAPQPCGC | |
RUO | |
RAB4B | |
Wheat Germ (in vitro) | |
GST | |
Liquid |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction