Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abnova™ Human RAD21 Full-length ORF (NP_006256.1, 1 a.a. - 631 a.a.) Recombinant Protein with GST-tag at N-terminal

Used for AP, Array, ELISA, WB-Re

Manufacturer:  Abnova™ H00005885P01

Catalog No. 89-959-237


Add to cart

Description

Description

The protein encoded by this gene is highly similar to the gene product of Schizosaccharomyces pombe rad21, a gene involved in the repair of DNA double-strand breaks, as well as in chromatid cohesion during mitosis. This protein is a nuclear phospho-protein, which becomes hyperphosphorylated in cell cycle M phase. The highly regulated association of this protein with mitotic chromatin specifically at the centromere region suggests its role in sister chromatid cohesion in mitotic cells. [provided by RefSeq]

Sequence: MFYAHFVLSKRGPLAKIWLAAHWDKKLTKAHVFECNLESSVESIISPKVKMALRTSGHLLLGVVRIYHRKAKYLLADCNEAFIKIKMAFRPGVVDLPEENREAAYNAITLPEEFHDFDQPLPDLDDIDVAQQFSLNQSRVEEITMREEVGNISILQENDFGDFGMDDREIMREGSAFEDDDMLVSTTTSNLLLESEQSTSNLNEKINHLEYEDQYKDDNFGEGNDGGILDDKLISNNDGGIFDDPPALSEAGVMLPEQPAHDDMDEDDNVSMGGPDSPDSVDPVEPMPTMTDQTTLVPNEEEAFALEPIDITVKETKAKRKRKLIVDSVKELDSKTIRAQLSDYSDIVTTLDLAPPTKKLMMWKETGGVEKLFSLPAQPLWNNRLLKLFTRCLTPLVPEDLRKRRKGGEADNLDEFLKEFENPEVPREDQQQQHQQRDVIDEPIIEEPSRLQESVMEASRTNIDESAMPPPPPQGVKRKAGQIDPEPVMPPQQVEQMEIPPVELPPEEPPNICQLIPELELLPEKEKEKEKEKEDDEEEEDEDASGGDQDQEERRWNKRTQQMLHGLQRALAKTGAESISLLELCRNTNRKQAAAKFYSFLVLKKQQAIELTQEEPYSDIIATPGPRFHII
Specifications

Specifications

NP_006256.1
Antibody Production, Array, Enzyme-linked Immunoabsorbent Assay, Western Blot (Recombinant protein)
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
98.1kDa
Glutathione Sepharose 4 Fast Flow
2 ug
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
FLJ25655|FLJ40596|HR21|HRAD21|KIAA0078|MCD1|NXP1|SCC1|hHR21
RAD21
Yes
wheat germ expression system
Liquid
5885
RAD21 (Human) Recombinant Protein (P01)
12.5% SDS-PAGE Stained with Coomassie Blue.
MFYAHFVLSKRGPLAKIWLAAHWDKKLTKAHVFECNLESSVESIISPKVKMALRTSGHLLLGVVRIYHRKAKYLLADCNEAFIKIKMAFRPGVVDLPEENREAAYNAITLPEEFHDFDQPLPDLDDIDVAQQFSLNQSRVEEITMREEVGNISILQENDFGDFGMDDREIMREGSAFEDDDMLVSTTTSNLLLESEQSTSNLNEKINHLEYEDQYKDDNFGEGNDGGILDDKLISNNDGGIFDDPPALSEAGVMLPEQPAHDDMDEDDNVSMGGPDSPDSVDPVEPMPTMTDQTTLVPNEEEAFALEPIDITVKETKAKRKRKLIVDSVKELDSKTIRAQLSDYSDIVTTLDLAPPTKKLMMWKETGGVEKLFSLPAQPLWNNRLLKLFTRCLTPLVPEDLRKRRKGGEADNLDEFLKEFENPEVPREDQQQQHQQRDVIDEPIIEEPSRLQESVMEASRTNIDESAMPPPPPQGVKRKAGQIDPEPVMPPQQVEQMEIPPVELPPEEPPNICQLIPELELLPEKEKEKEKEKEDDEEEEDEDASGGDQDQEERRWNKRTQQMLHGLQRALAKTGAESISLLELCRNTNRKQAAAKFYSFLVLKKQQAIELTQEEPYSDIIATPGPRFHII
RUO
RAD21
Wheat Germ (in vitro)
GST
SDS
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit