Learn More
Abnova™ Human RAD54B Full-length ORF (AAH33710.2, 1 a.a. - 158 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
Supplier: Abnova™ H00025788P01S
Description
The protein encoded by this gene belongs to the DEAD-like helicase superfamily. It shares similarity with Saccharomyces cerevisiae RAD54 and RDH54, both of which are involved in homologous recombination and repair of DNA. This protein binds to double-stranded DNA, and displays ATPase activity in the presence of DNA. This gene is highly expressed in testis and spleen, which suggests active roles in meiotic and mitotic recombination. Homozygous mutations of this gene were observed in primary lymphoma and colon cancer. [provided by RefSeq]
Sequence: MLDHHPVAITVEVKQEEDIKPPPPLVLNSQQSDTLEQREEHELVHVMERSLSPSLSSVDMRMTSSPSSIPRRDDFFRHESGEHFRSLLGYDPQILQMLKEEHQIILENQKNFGLYVQEKRDGLKRRQQLEEELLRAKIEVEKLKAIRLRHDLPEYNSLSpecifications
AAH33710.2 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
45kDa | |
Glutathione Sepharose 4 Fast Flow | |
10 ug | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
FSBP/RDH54 | |
RAD54B | |
Recombinant | |
wheat germ expression system |
Antibody Production, Protein Array, ELISA, Western Blot | |
25788 | |
RAD54B (Human) Recombinant Protein (P01) | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
MLDHHPVAITVEVKQEEDIKPPPPLVLNSQQSDTLEQREEHELVHVMERSLSPSLSSVDMRMTSSPSSIPRRDDFFRHESGEHFRSLLGYDPQILQMLKEEHQIILENQKNFGLYVQEKRDGLKRRQQLEEELLRAKIEVEKLKAIRLRHDLPEYNSL | |
RUO | |
RAD54B | |
Wheat Germ (in vitro) | |
GST | |
Liquid |
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.