Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Eisco™ Human Radius Bone Model

Life-size human radius bone model provides an incredible amount of realism and detail for examining the human skeleton
$11.50
Specifications
For Use With (Application) | Examining The Human Skeleton |
---|---|
Length (English) | 9 in. |
Material | PVC |
Type | Anatomical Model, Human |
Width (English) | 1.25 in. |
Description
- Life-size, anatomically correct replica
- Realistic details, textures, and bony landmarks
- Made of durable and sturdy PVC plastic
- Ideal for both student and patient educational demonstrations
- Washable
Specifications
Examining The Human Skeleton | |
PVC | |
1.25 in. |
9 in. | |
Anatomical Model, Human |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title