Learn More
Abnova™ Human RASA3 Full-length ORF (NP_031394.2, 1 a.a. - 834 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
$499.00 - $769.00
Specifications
Accession Number | NP_031394.2 |
---|---|
For Use With (Application) | Antibody Production, Protein Array, ELISA, Western Blot |
Formulation | 50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. |
Gene ID (Entrez) | 22821 |
Molecular Weight (g/mol) | 122.1kDa |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
89-959-425
|
Abnova™
H00022821P01L |
25 ug |
Each for $769.00
|
|
|||||
89-959-424
|
Abnova™
H00022821P01S |
10 ug |
Each for $499.00
|
|
|||||
Description
The protein encoded by this gene is member of the GAP1 family of GTPase-activating proteins. The gene product stimulates the GTPase activity of normal RAS p21 but not its oncogenic counterpart. Acting as a suppressor of RAS function, the protein enhances the weak intrinsic GTPase activity of RAS proteins resulting in the inactive GDP-bound form of RAS, thereby allowing control of cellular proliferation and differentiation. This family member is an inositol 1,3,4,5-tetrakisphosphate-binding protein, like the closely related RAS p21 protein activator 2. The two family members have distinct pleckstrin-homology domains, with this particular member having a domain consistent with its localization to the plasma membrane. [provided by RefSeq]
Sequence: MAVEDEGLRVFQSVKIKIGEAKNLPSYPGPSKMRDCYCTVNLDQEEVFRTKIVEKSLCPFYGEDFYCEIPRSFRHLSFYIFDRDVFRRDSIIGKVAIQKEDLQKYHNRDTWFQLQHVDADSEVQGKVHLELRLSEVITDTGVVCHKLATRIVECQGLPIVNGQCDPYATVTLAGPFRSEAKKTKVKRKTNNPQFDEVFYFEVTRPCSYSKKSHFDFEEEDVDKLEIRVDLWNASNLKFGDEFLGELRIPLKVLRQSSSYEAWYFLQPRDNGSKSLKPDDLGSLRLNVVYTEDHVFSSDYYSPLRDLLLKSADVEPVSASAAHILGEVCREKQEAAVPLVRLFLHYGRVVPFISAIASAEVKRTQDPNTIFRGNSLASKCIDETMKLAGMHYLHVTLKPAIEEICQSHKPCEIDPVKLKDGENLENNMENLRQYVDRVFHAITESGVSCPTVMCDIFFSLREAAAKRFQDDPDVRYTAVSSFIFLRFFAPAILSPNLFQLTPHHTDPQTSRTLTLISKTVQTLGSLSKSKSASFKESYMATFYEFFNEQKYADAVKNFLDLISSSGRRDPKSVEQPIVLKEGFMIKRAQGRKRFGMKNFKKRWFRLTNHEFTYHKSKGDQPLYSIPIENILAVEKLEEESFKMKNMFQVIQPERALYIQANNCVEAKDWIDILTKVSQCNQKRLTVYHPSAYLSGHWLCCRAPSDSAPGCSPCTGGLPANIQLDIDGDRETERIYSLFNLYMSKLEKMQEACGSKSVYDGPEQEEYSTFVIDDPQETYKTLKQVIAGVGALEQEHAQYKRDKFKKTKYGSQEHPIGDKSFQNYIRQQSETSTHSISpecifications
NP_031394.2 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
122.1kDa | |
Glutathione Sepharose 4 Fast Flow | |
MAVEDEGLRVFQSVKIKIGEAKNLPSYPGPSKMRDCYCTVNLDQEEVFRTKIVEKSLCPFYGEDFYCEIPRSFRHLSFYIFDRDVFRRDSIIGKVAIQKEDLQKYHNRDTWFQLQHVDADSEVQGKVHLELRLSEVITDTGVVCHKLATRIVECQGLPIVNGQCDPYATVTLAGPFRSEAKKTKVKRKTNNPQFDEVFYFEVTRPCSYSKKSHFDFEEEDVDKLEIRVDLWNASNLKFGDEFLGELRIPLKVLRQSSSYEAWYFLQPRDNGSKSLKPDDLGSLRLNVVYTEDHVFSSDYYSPLRDLLLKSADVEPVSASAAHILGEVCREKQEAAVPLVRLFLHYGRVVPFISAIASAEVKRTQDPNTIFRGNSLASKCIDETMKLAGMHYLHVTLKPAIEEICQSHKPCEIDPVKLKDGENLENNMENLRQYVDRVFHAITESGVSCPTVMCDIFFSLREAAAKRFQDDPDVRYTAVSSFIFLRFFAPAILSPNLFQLTPHHTDPQTSRTLTLISKTVQTLGSLSKSKSASFKESYMATFYEFFNEQKYADAVKNFLDLISSSGRRDPKSVEQPIVLKEGFMIKRAQGRKRFGMKNFKKRWFRLTNHEFTYHKSKGDQPLYSIPIENILAVEKLEEESFKMKNMFQVIQPERALYIQANNCVEAKDWIDILTKVSQCNQKRLTVYHPSAYLSGHWLCCRAPSDSAPGCSPCTGGLPANIQLDIDGDRETERIYSLFNLYMSKLEKMQEACGSKSVYDGPEQEEYSTFVIDDPQETYKTLKQVIAGVGALEQEHAQYKRDKFKKTKYGSQEHPIGDKSFQNYIRQQSETSTHSI | |
RUO | |
RASA3 | |
Wheat Germ (in vitro) | |
GST | |
Liquid |
Antibody Production, Protein Array, ELISA, Western Blot | |
22821 | |
RASA3 (Human) Recombinant Protein (P01) | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
GAP1IP4BP/GAPIII/MGC46517/MGC47588 | |
RASA3 | |
Recombinant | |
wheat germ expression system |
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.