Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Abnova™ Human RBM12 Partial ORF (NP_006038.2, 834 a.a. - 932 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
Supplier: Abnova™ H00010137Q01S
Description
This gene encodes a protein that contains several RNA-binding motifs, potential transmembrane domains, and proline-rich regions. This gene and the gene for copine I overlap at map location 20q11.21. Alternative splicing in the 5' UTR results in two transcript variants. Both variants encode the same protein. [provided by RefSeq]
Sequence: GPGPIHIGGPPGFASSSGKPGPTVIKVQNMPFTVSIDEILDFFYGYQVIPGSVCLKYNEKGMPTGEAMVAFESRDEATAAVIDLNDRPIGSRKVKLVLGSpecifications
NP_006038.2 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
36.63kDa | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
GPGPIHIGGPPGFASSSGKPGPTVIKVQNMPFTVSIDEILDFFYGYQVIPGSVCLKYNEKGMPTGEAMVAFESRDEATAAVIDLNDRPIGSRKVKLVLG | |
RUO | |
RBM12 | |
Wheat Germ (in vitro) | |
GST | |
Liquid |
Antibody Production, ELISA, Protein Array, Western Blot | |
10137 | |
RBM12 (Human) Recombinant Protein (Q01) | |
10 ug | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
HRIHFB2091/KIAA0765/SWAN | |
RBM12 | |
Recombinant | |
wheat germ expression system |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction