Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abnova™ Human RCHY1 Full-length ORF (NP_001009922.1, 1 a.a. - 252 a.a.) Recombinant Protein with GST-tag at N-terminal

Used for AP, Array, ELISA, WB-Re

Supplier:  Abnova™ H00025898P01S

 View more versions of this product

Catalog No. 89-959-648


Add to Cart

Description

Description

The protein encoded by this gene has ubiquitin-protein ligase activity. This protein binds with p53 and promotes the ubiquitin-mediated proteosomal degradation of p53. This gene is oncogenic because loss of p53 function contributes directly to malignant tumor development. Transcription of this gene is regulated by p53. Alternative splicing results in multiple transcript variants encoding different isoforms. [provided by RefSeq]

Sequence: MAATAREDGASGQERGQRGCEHYDRGCLLKAPCCDKLYTCRLCHDNNEDHQLDRFKVKEVQCINCEKIQHAQQTCEECSTLFGEYYCDICHLFDKDKKQYHCENCGICRIGPKEDFFHCLKCNLCLAMNLQGRHKCIENVSRQNCPICLEDIHTSRVVAHVLPCGHLLHRGYRCPLCMHSALDMTRYWRQLDDEVAQTPMPSEYQNMTVDILCNDCNGRSTVQFHILGMKCKICESYNTAQAGGRRISLDQQ
Specifications

Specifications

NP_001009922.1
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
55.4kDa
Glutathione Sepharose 4 Fast Flow
10 ug
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
ARNIP/CHIMP/DKFZp586C1620/PIRH2/PRO1996/RNF199/ZNF363/hARNIP
RCHY1
Recombinant
wheat germ expression system
Antibody Production, Protein Array, ELISA, Western Blot
25898
RCHY1 (Human) Recombinant Protein (P01)
12.5% SDS-PAGE Stained with Coomassie Blue.
MAATAREDGASGQERGQRGCEHYDRGCLLKAPCCDKLYTCRLCHDNNEDHQLDRFKVKEVQCINCEKIQHAQQTCEECSTLFGEYYCDICHLFDKDKKQYHCENCGICRIGPKEDFFHCLKCNLCLAMNLQGRHKCIENVSRQNCPICLEDIHTSRVVAHVLPCGHLLHRGYRCPLCMHSALDMTRYWRQLDDEVAQTPMPSEYQNMTVDILCNDCNGRSTVQFHILGMKCKICESYNTAQAGGRRISLDQQ
RUO
RCHY1
Wheat Germ (in vitro)
GST
Liquid
Product Suggestions

Product Suggestions

SDS
Documents

Documents

Product Certifications
Promotions

Promotions

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit