Learn More
Abnova Human RDH5 Partial ORF (AAH28298, 181 a.a. - 280 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
Manufacturer: Abnova H00005959Q01L
Description
This gene encodes an enzyme belonging to the short-chain dehydrogenases/reductases (SDR) family. This retinol dehydrogenase functions to catalyze the final step in the biosynthesis of 11-cis retinaldehyde, which is the universal chromophore of visual pigments. Mutations in this gene cause autosomal recessive fundus albipunctatus, a rare form of night blindness that is characterized by a delay in the regeneration of cone and rod photopigments. [provided by RefSeq]
Sequence: GLEAFSDSLRRDVAHFGIRVSIVEPGFFRTPVTNLESLEKTLQACWARLPPATQAHYGGAFLTKYLKMQQRIMNLICDPDLTKVSRCLEHALTARHPRTRSpecifications
AAH28298 | |
Antibody Production, Enzyme-linked Immunoabsorbent Assay, Protein Array, Western Blot (Recombinant protein) | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
36.63kDa | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
GLEAFSDSLRRDVAHFGIRVSIVEPGFFRTPVTNLESLEKTLQACWARLPPATQAHYGGAFLTKYLKMQQRIMNLICDPDLTKVSRCLEHALTARHPRTR | |
RUO | |
RDH5 | |
Wheat Germ (in vitro) | |
GST |
wheat germ expression system | |
Liquid | |
5959 | |
RDH5 (Human) Recombinant Protein (Q01) | |
25 ug | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
FLJ39337|FLJ97089|HSD17B9|RDH1|SDR9C5 | |
RDH5 | |
Yes |
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.