Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abnova™ Human RDH8 Full-length ORF (AAI46480.1, 1 a.a. - 311 a.a.) Recombinant Protein with GST-tag at N-terminal

Used for AP, Array, ELISA, WB-Re

Manufacturer:  Abnova™ H00050700P01S

 View more versions of this product

Catalog No. 89-959-689


Add to cart

Description

Description

All-trans-retinol dehydrogenase (RDH8) is a visual cycle enzyme that reduces all-trans-retinal to all-trans-retinol in the presence of NADPH (Rattner et al., 2000 [PubMed 10753906]). It is a member of the short chain dehydrogenase/reductase family and is located in the outer segments of photoreceptors; hence it is also known as photoreceptor retinol dehydrogenase. It is important in the visual cycle by beginning the rhodopsin regeneration pathway by reducing all-trans-retinal, the product of bleached and hydrolysed rhodopsin (Rando, 2001 [PubMed 11710234]). This is a rate-limiting step in the visual cycle (Saari et al., 1998 [PubMed 9667000]).[supplied by OMIM]

Sequence: MAAAPRTVLISGCSSGIGLELAVQLAHDPKKRYQVVATMRDLGKKETLEAAAGEALGQTLTVAQLDVCSDESVAQCLSCIQGEVDVLVNNAGMGLVGPLEGLSLAAMQNVFDTNFFGAVRLVKAVLPGMKRRRQGHIVVISSVMGLQGVIFNDVYAASKFALEGFFESLAIQLLQFNIFISLVEPGPVVTEFEGKLLAQVSMAEFPGTDPETLHYFRDLYLPASRKLFCSVGQNPQDVVQAIVNVISSTRPPLRRQTNIRYSPLTTLKTVDSSGSLYVRTTHRLLFRCPRLLNLGLQCLSCGCLPTRVRPR
Specifications

Specifications

AAI46480.1
Antibody Production, Array, Enzyme-linked Immunoabsorbent Assay, Western Blot (Recombinant protein)
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
61.16kDa
Glutathione Sepharose 4 Fast Flow
10 ug
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
PRRDH|SDR28C2
RDH8
Yes
wheat germ expression system
Liquid
50700
RDH8 (Human) Recombinant Protein (P01)
12.5% SDS-PAGE Stained with Coomassie Blue.
MAAAPRTVLISGCSSGIGLELAVQLAHDPKKRYQVVATMRDLGKKETLEAAAGEALGQTLTVAQLDVCSDESVAQCLSCIQGEVDVLVNNAGMGLVGPLEGLSLAAMQNVFDTNFFGAVRLVKAVLPGMKRRRQGHIVVISSVMGLQGVIFNDVYAASKFALEGFFESLAIQLLQFNIFISLVEPGPVVTEFEGKLLAQVSMAEFPGTDPETLHYFRDLYLPASRKLFCSVGQNPQDVVQAIVNVISSTRPPLRRQTNIRYSPLTTLKTVDSSGSLYVRTTHRLLFRCPRLLNLGLQCLSCGCLPTRVRPR
RUO
RDH8
Wheat Germ (in vitro)
GST
SDS
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit