Learn More
STEMCELL Technologies Human Recombinant IFN-beta, Size: 5 µg
SDP
Supplier:  STEMCELL Technologies 78113
Encompass_Preferred
Interferon beta (IFN-β), also known as type I interferon, is produced by fibroblasts, and in smaller amounts by plasmocytoid dendritic cells. Macrophages and endothelial cells secrete IFN-β in response to a viral infection (Reder & Feng). IFN-β binds to a receptor complex composed of IFNAR1 and IFNAR2, and initiates signal transduction via the JAK/STAT pathway; this culminates in the transcription and activation of many genes that control dendritic cell activation, T cell survival, NK cell activation, chemokine expression, lymph node retention, and antiproliferative and antiviral effects (Dunn et al.). IFN-β is a first-line treatment for multiple sclerosis. It suppresses Th17 cells by affecting expression of IL-4, IL-10, and IL-27. IFN-β was also shown to expand regulatory T cells and limit T cell trafficking to the central nervous system (Inoue & Shinohara). Out of the two variants of IFN-β (IFN-β1 and IFN-β3), this product is the IFN-β1 form.
Catalog No. 50-197-6496
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.