Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human Reduced Folate Carrier/SLC19A1 Antibody, R&D Systems™

Mouse Monoclonal Antibody
Supplier: R&D Systems MAB8450
This item is not returnable.
View return policy
Description
Reduced Folate Carrier/SLC19A1 Monoclonal specifically detects Reduced Folate Carrier/SLC19A1 in Human samples. It is validated for Flow Cytometry, CyTOF-ready.Specifications
Reduced Folate Carrier/SLC19A1 | |
Monoclonal | |
Unconjugated | |
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
CHMD, FLOT1, folate transporter 1, FOLT, FOLThuman reduced folate carrier (RFC)10Intestinal folate carrier 1, IFC1, IFC-1, Placental folate transporter, Reduced folate carrier protein, REFC, RFC, RFC1, solute carrier family 19 (folate transporter), member 1, Solute carrier family 19 member 1 | |
Mouse | |
Protein A or G purified from hybridoma culture supernatant | |
RUO | |
6573 | |
Reconstitute at 0.5 mg/mL in sterile PBS. | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
IgG2a |
Flow Cytometry, CyTOF | |
890513 | |
Flow Cytometry 0.25 ug/10^6 cells, CyTOF-ready | |
P41440 | |
SLC19A1 | |
NS0 mouse myeloma cell line transfected with human Reduced Folate Carrier/SLC19A1 Met1-Gln591 Accession # P41440 | |
100 μg | |
Primary | |
Stains human Reduced Folate Carrier/SLC19A1 transfectants but not irrelevant transfectants in flow cytometry. | |
Human | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction