Learn More
Abnova Human REG1B Partial ORF (NP_006498.1, 67 a.a. - 166 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
Manufacturer: Abnova H00005968Q01S
Description
This gene is a type I subclass member of the Reg gene family. The Reg gene family is a multigene family grouped into four subclasses, types I, II, III and IV based on the primary structures of the encoded proteins. This gene encodes a protein secreted by the exocrine pancreas that is highly similar to the REG1A protein. The related REG1A protein is associated with islet cell regeneration and diabetogenesis, and may be involved in pancreatic lithogenesis. Reg family members REG1A, REGL, PAP and this gene are tandemly clustered on chromosome 2p12 and may have arisen from the same ancestral gene by gene duplication. [provided by RefSeq]
Sequence: MNSGNLVSVLTQAEGAFVASLIKESSTDDSNVWIGLHDPKKNRRWHWSSGSLVSYKSWDTGSPSSANAGYCASLTSCSGFKKWKDESCEKKFSFVCKFKNSpecifications
NP_006498.1 | |
Antibody Production, Enzyme-linked Immunoabsorbent Assay, Protein Array, Western Blot (Recombinant protein) | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
36.74kDa | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
MNSGNLVSVLTQAEGAFVASLIKESSTDDSNVWIGLHDPKKNRRWHWSSGSLVSYKSWDTGSPSSANAGYCASLTSCSGFKKWKDESCEKKFSFVCKFKN | |
RUO | |
REG1B | |
Wheat Germ (in vitro) | |
GST |
wheat germ expression system | |
Liquid | |
5968 | |
REG1B (Human) Recombinant Protein (Q01) | |
10 ug | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
PSPS2|REGH|REGI-BETA|REGL | |
REG1B | |
Yes |
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.