Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Human Reg3A ELISA Kit

Catalog No. p-7219596
Encompass_Preferred
Click to view available options
Quantity:
5 x 96 Tests
96 Tests

ELISA

Human Reg3A quantitates human Reg3A in serum, plasma, supernatant. The assay will exclusively recognize both natural and recombinant human Reg3A.

REG3A encodes a pancreatic secretory protein that may be involved in cell proliferation or differentiation. It has similarity to the C-type lectin superfamily. The enhanced expression of this protein is observed during pancreatic inflammation and liver carcinogenesis.
TRUSTED_SUSTAINABILITY

Specifications

Accession Number AV063448; generating islet-derived, mouse homolog 3 alpha; hepatocarcinoma-intestine-pancreas; hepatointestinal pancreatic protein; HIP; HIP/PAP; Human proislet peptide; INGAP; Islet of Langerhans regenerating protein 3; lithostathine 3; pancreatic beta cell growth factor; pancreatitis-associated protein 1; Pancreatitis-associated protein 2; PAP; PAP homologous protein; PAP1; Pap2; PAP-H; PapII; PBCGF; proliferation-inducing protein 34; proliferation-inducing protein 42; REG 3; REG 3 A; REG 3 alpha; Reg IIIalpha; Reg III-alpha; REG3; REG3 alpha; Reg3a; REG3alpha; REG-3 alpha; REG-3-alpha; regeneratin; regenerating family member 3 alpha; regenerating islet-derived 3 alpha; regenerating islet-derived protein 3 alpha; regenerating islet-derived protein 3-alpha; Regenerating islet-derived protein 3-alpha 15 kDa form; Regenerating islet-derived protein 3-alpha 16.5 kDa form; Regenerating islet-derived protein III-alpha; regIII; REG-III; RegIII (alpha)
Assay Range 0.12 to 25 ng/mL
Assay Sensitivity 0.12 ng/mL
Conjugate HRP
Product Type ELISA
Sample Type Plasma, Serum, Supernatant
For Use With (Equipment) Colorimetric Microplate Reader
Gene ID (Entrez) 5068
Gene Symbol Reg3a
Interassay CV <12%
Intraassay CV <10
Kit Contents Pre-coated 96 well plate, Standard, Assay Diluent concentrate, Biotinylated Detection Antibody, SAV-HRP, Wash Buffer, Chromogen, Stop Solution, Adhesive Plate Covers
Label or Dye HRP
Quantity 5 x 96 Tests
Regulatory Status RUO
Sample Volume Plasma 50 μL, Serum 50 μL, Supernatant 50 μL
Storage Requirements 2°C to 8°C
Target REG3A
Test Time 1 hrs 20 mins
Total Assay Time 4 hrs 45 mins
Show More Show Less
WARNING: Reproductive Harm - www.P65Warnings.ca.gov
Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.