Learn More
Raybiotech IncSupplier Diversity Partner Human Renin ELISA Kit DFS Item
Manufacturer:  Raybiotech IncSupplier Diversity Partner ELHRENIN1
For serum, plasma, and cell culture supernatant samples. Sensitivity: 0.41 ng/ml. Pre-coated 96-well strip plate with additional reagents to allow for use in multiple experiments. Specificity: Cross Reactivity: This ELISA kit shows no cross-reactivity with the following cytokines tested: human ADAMTS-13, Aggrecan, Angiotensinogen, B7-H1, BMPR-IA (ALK-3), BMPR-II, Cadherin-11, CD27 (TNFRSF7), CD6, Ck beta 8-1 (CCL23), CNTF, DNAM- 1 (CD226), CD147 (EMMPRIN), FLRG, Follistatin-like 1, Fractalkine, Galectin-1, GITR Ligand, Granulysin (NKG5/LAG-2), IL-1 R3 (IL-1 R Acp), IL-15 R alpha, IL-17E (IL-25), IL-32 alpha, CHL-1 (L1CAM-2), LRIG3, LRP-6, MEPE (OF45), Nectin-4, Periostin, Persephin, RGM-B, ROBO3, S100 A8, Siglec-7, Syndecan-3, Thrombospondin 2, Thrombospondin 5, Tie-1, ULBP-2.
Catalog No. 50-194-9683