Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Gibco™ Human Resistin Recombinant Protein, PeproTech®
E. coli Recombinant Protein
$89.50 - $5770.00
Specifications
Accession Number | Q9HD89 |
---|---|
For Use With (Application) | ELISA, Functional Assay, Western Blot |
Formulation | protein with no preservative |
Gene ID (Entrez) | 56729 |
Molecular Weight (g/mol) | 19.5 |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
45019100UG
|
Gibco™
45019100UG |
100 μg |
Each for $776.00
|
|
|||||
450-19-1MG
|
Gibco™
450191MG |
1 mg |
Each for $5,770.00
|
|
|||||
45019250UG
|
Gibco™
45019250UG |
250 μg |
Each for $1,942.00
|
|
|||||
450-19-25UG
|
Gibco™
4501925UG |
25 μg |
Each for $217.00
|
|
|||||
45019500UG
|
Gibco™
45019500UG |
500 μg |
Each for $3,460.00
|
|
|||||
450-19-50UG
|
Gibco™
4501950UG |
50 μg |
Each for $400.00
|
|
|||||
450-19-5UG
|
Gibco™
450195UG |
5 μg |
Each for $89.50
|
|
|||||
Description
Adipose tissue is an energy reserve in animals and is strictly regulated. Adipose cells produce and secrete numerous physiologically important proteins, such as lipoprotein lipase (LPL), leptin, adipocyte complement related protein of 30 kDa (Acrp30), perilipin, and resistin. Resistin, also known as Adipocyte Secreted Factor (ADSF) and FIZZ3, is a cytokine that is specifically secreted by adipocytes. While the function of resistin is not well understood, it appears to have key paracrine roles and has a suggested role in type 2 diabetes. Studies show that in obese, diabetic mice the levels of serum resistin are increased, which has an antagonistic effect on insulin. Additional studies have shown that resistin causes insulin resistance and glucose intolerance in mice. Resistin gene expression requires C/EBP alpha, which is necessary and sufficient for expression. Studies show that the coactivators CBP and p300, which have been shown to display histone acetyltransferase (HAT) activity, abundantly acetylate the histones at the resistin promoter suggesting their role in activating chromatin for resistin transcription. Numerous PPAR gamma and RXR ligands have been shown to downregulate resistin expression in adipocytes.Specifications
Q9HD89 | |
protein with no preservative | |
19.5 | |
E. coli | |
RUO | |
Adipose tissue specific secretory factor; adipose tissue-specific secretory factor; Adipose-specific cysteine-rich secreted protein A12-alpha; ADSF; C/EBP epsilon regulated myeloid specific secreted cysteine rich protein precursor 1; C/EBP-epsilon regulated myeloid-specific secreted cysteine-rich protein precursor 1; c/EBP-epsilon-regulated myeloid-specific secreted cysteine-rich protein; cysteine-r; Cysteine-rich secreted protein A12-alpha-like 2; cysteine-rich secreted protein FIZZ3; dominant inhibitory adipocyte-specific secretory factor; dri; FIZZ3; found in inflammatory zone 3; HXCP1; MGC126603; MGC126609; Resistin; resistin delta2; RETN; RETN1; RSN; RSTN; UNQ407/PRO1199; XCP1; Xcp4 | |
Human | |
Recombinant | |
Lyophilized |
ELISA, Functional Assay, Western Blot | |
56729 | |
Human Resistin | |
-20°C | |
<1 EU/μg | |
RETN | |
Unconjugated | |
SSKTLCSMEE AINERIQEVA GSLIFRAISS IGLECQSVTS RGDLATCPRG FAVTGCTCGS ACGSWDVRAE TTCHCQCAGM DWTGARCCRV QP | |
≥ 98% by SDS-PAGE gel and HPLC analyses. |
Spot an opportunity for improvement?
Provide Content Correction
Provide Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title