Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human Respiratory Syncytial Virus A2 Fusion Protein Antibody, DyLight 405, Novus Biologicals™
Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP326093V
Description
Human Respiratory Syncytial Virus A2 Fusion Protein Polyclonal antibody specifically detects Human Respiratory Syncytial Virus A2 Fusion Protein in Virus samples. It is validated for Western BlotSpecifications
Human Respiratory Syncytial Virus A2 Fusion Protein | |
Polyclonal | |
Western Blot | |
HRSVgp08, RSV, RSV fusion protein | |
Rabbits immunized with purified, recombinant Human Respiratory Syncytial Virus A2 Fusion Protein (Uniprot#: P03420.1, Met1-Thr529) | |
0.1 mL | |
Primary | |
Store at 4C in the dark. | |
IgG |
Western Blot | |
DyLight 405 | |
50mM Sodium Borate | |
Rabbit | |
Antigen and protein A Affinity-purified | |
RUO | |
Virus | |
Purified |
Provide Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction