Learn More
Bon Opus Biosciences
Recombinant Human Fibronectin Fragment/RetroNectin/NovoNectin
Manufacturer: Bon Opus Biosciences CH3810UG
Recombinant Human Fibronectin fragment is produced by our E.coli expression system and the target gene encoding Pro1270-Ser1546&Ala1721-Thr2016 is expressed. Bon Opus Cat. #CH38