Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Abnova™ Human RFFL Full-length ORF (AAH28424.1, 1 a.a. - 327 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
$416.00 - $641.00
Specifications
Name | RFFL (Human) Recombinant Protein (P01) |
---|---|
Recombinant | Yes |
Purification Method | Glutathione Sepharose 4 Fast Flow |
For Use With (Application) | Antibody Production, Array, Enzyme-linked Immunoabsorbent Assay, Western Blot (Recombinant protein) |
Molecular Weight | 63kDa |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
89-959-801
|
Abnova™
H00117584P01L |
25 ug |
Each for $641.00
|
|
89-959-800
|
Abnova™
H00117584P01S |
10 ug |
Each for $416.00
|
|
Description
Sequence: MWATCCNWFCLDGQPEEVPPPQGARMQAYSNPGYSSFPSPTGLEPSCKSCGAHFANTARKQTCLDCKKNFCMTCSSQVGNGPRLCLLCQRFRATAFQREELMKMKVKDLRDYLSLHDISTEMCREKGELVLLVLGQQPVISQEDRTRASTLSPDFPEQQAFLTQPHSSMVPPTSPNLPSSSAQATSVPPAQVQENQQSIDSEDSFVPGRRASLSDLTDLEDIEGLTVRQLKEILARNFVNYKGCCEKWELMERVTRLYKDQKGLQHLGGAVPSGLEENLCKICMDSPIDCVLLECGHMVTCTKCGKRMNECPICRQYVIRAVHVFRSSpecifications
RFFL (Human) Recombinant Protein (P01) | |
Glutathione Sepharose 4 Fast Flow | |
63kDa |
Yes | |
Antibody Production, Array, Enzyme-linked Immunoabsorbent Assay, Western Blot (Recombinant protein) | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title