Learn More
Abnova Human RFX2 Partial ORF (NP_000626, 323 a.a. - 429 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
Manufacturer: Abnova H00005990Q01L
Description
This gene is a member of the regulatory factor X gene family, which encodes transcription factors that contain a highly-conserved winged helix DNA binding domain. The protein encoded by this gene is structurally related to regulatory factors X1, X3, X4, and X5. It is a transcriptional activator that can bind DNA as a monomer or as a heterodimer with other RFX family members. This protein can bind to cis elements in the promoter of the IL-5 receptor alpha gene. Two transcript variants encoding different isoforms have been described for this gene, and both variants utilize alternative polyadenylation sites. [provided by RefSeq]
Sequence: PEQTMAVQSQHHQQYIDVSHVFPEFPAPDLGSFLLQDGVTLHDVKALQLVYRRHCEATVDVVMNLQFHYIEKLWLSFWNSKASSSDGPTSLPASDEDPEGAVLPKDKSpecifications
NP_000626 | |
Antibody Production, Enzyme-linked Immunoabsorbent Assay, Protein Array, Western Blot (Recombinant protein) | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
37.51kDa | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
PEQTMAVQSQHHQQYIDVSHVFPEFPAPDLGSFLLQDGVTLHDVKALQLVYRRHCEATVDVVMNLQFHYIEKLWLSFWNSKASSSDGPTSLPASDEDPEGAVLPKDK | |
RUO | |
RFX2 | |
Wheat Germ (in vitro) | |
GST |
wheat germ expression system | |
Liquid | |
5990 | |
RFX2 (Human) Recombinant Protein (Q01) | |
25 ug | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
FLJ14226 | |
RFX2 | |
Yes |
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.