Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abnova™ Human RFX4 Full-length ORF (NP_115880.2, 1 a.a. - 641 a.a.) Recombinant Protein with GST-tag at N-terminal

Used for AP, Array, ELISA, WB-Re

Manufacturer:  Abnova™ H00005992P02S

 View more versions of this product

Catalog No. 89-959-838


Add to cart

Description

Description

This gene is a member of the regulatory factor X gene family, which encodes transcription factors that contain a highly-conserved winged helix DNA binding domain. The protein encoded by this gene is structurally related to regulatory factors X1, X2, X3, and X5. It has been shown to interact with itself as well as with regulatory factors X2 and X3, but it does not interact with regulatory factor X1. This protein may be a transcriptional repressor rather than a transcriptional activator. Three transcript variants encoding different isoforms have been described for this gene. [provided by RefSeq]

Sequence: MNWAAFGGSEFFIPEGIQIDSRCPLSRNITEWYHYYGIAVKESSQYYDVMYSKKGAAWVSETGKKEVSKQTVAYSPRSKLGTLLPEFPNVKDLNLPASLPEEKVSTFIMMYRTHCQRILDTVIRANFDEVQSFLLHFWQGMPPHMLPVLGSSTVVNIVGVCDSILYKAISGVLMPTVLQALPDSLTQVIRKFAKQLDEWLKVALHDLPENLRNIKFELSRRFSQILRRQTSLNHLCQASRTVIHSADITFQMLEDWRNVDLNSITKQTLYTMEDSRDEHRKLITQLYQEFDHLLEEQSPIESYIEWLDTMVDRCVVKVAAKRQGSLKKVAQQFLLMWSCFGTRVIRDMTLHSAPSFGSFHLIHLMFDDYVLYLLESLHCQERANELMRAMKGEGSTAEVREEIILTEAAAPTPSPVPSFSPAKSATSVEVPPPSSPVSNPSPEYTGLSTTGAMQSYTWSLTYTVTTAAGSPAENSQQLPCMRNTHVPSSSVTHRIPVYPHREEHGYTGSYNYGSYGNQHPHPMQSQYPALPHDTAISGPLHYAPYHRSSAQYPFNSPTSRMEPCLMSSTPRLHPTPVTPRWPEVPSANTCYTSPSVHSARYGNSSDMYTPLTTRRNSEYEHMQHFPGFAYINGEASTGWAK
Specifications

Specifications

NP_115880.2
Antibody Production, Array, Enzyme-linked Immunoabsorbent Assay, Western Blot (Recombinant protein)
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
99kDa
Glutathione Sepharose 4 Fast Flow
10 ug
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
NYD-SP10
RFX4
Yes
wheat germ expression system
Liquid
5992
RFX4 (Human) Recombinant Protein (P02)
12.5% SDS-PAGE Stained with Coomassie Blue.
MNWAAFGGSEFFIPEGIQIDSRCPLSRNITEWYHYYGIAVKESSQYYDVMYSKKGAAWVSETGKKEVSKQTVAYSPRSKLGTLLPEFPNVKDLNLPASLPEEKVSTFIMMYRTHCQRILDTVIRANFDEVQSFLLHFWQGMPPHMLPVLGSSTVVNIVGVCDSILYKAISGVLMPTVLQALPDSLTQVIRKFAKQLDEWLKVALHDLPENLRNIKFELSRRFSQILRRQTSLNHLCQASRTVIHSADITFQMLEDWRNVDLNSITKQTLYTMEDSRDEHRKLITQLYQEFDHLLEEQSPIESYIEWLDTMVDRCVVKVAAKRQGSLKKVAQQFLLMWSCFGTRVIRDMTLHSAPSFGSFHLIHLMFDDYVLYLLESLHCQERANELMRAMKGEGSTAEVREEIILTEAAAPTPSPVPSFSPAKSATSVEVPPPSSPVSNPSPEYTGLSTTGAMQSYTWSLTYTVTTAAGSPAENSQQLPCMRNTHVPSSSVTHRIPVYPHREEHGYTGSYNYGSYGNQHPHPMQSQYPALPHDTAISGPLHYAPYHRSSAQYPFNSPTSRMEPCLMSSTPRLHPTPVTPRWPEVPSANTCYTSPSVHSARYGNSSDMYTPLTTRRNSEYEHMQHFPGFAYINGEASTGWAK
RUO
RFX4
Wheat Germ (in vitro)
GST
SDS
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit