Learn More
Abnova Human RFX5 Full-length ORF (NP_000440.1, 1 a.a. - 616 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
Manufacturer: Abnova H00005993P01L
Description
A lack of MHC-II expression results in a severe immunodeficiency syndrome called MHC-II deficiency, or the bare lymphocyte syndrome (BLS; MIM 209920). At least 4 complementation groups have been identified in B-cell lines established from patients with BLS. The molecular defects in complementation groups B, C, and D all lead to a deficiency in RFX, a nuclear protein complex that binds to the X box of MHC-II promoters. The lack of RFX binding activity in complementation group C results from mutations in the RFX5 gene encoding the 75-kD subunit of RFX (Steimle et al., 1995). RFX5 is the fifth member of the growing family of DNA-binding proteins sharing a novel and highly characteristic DNA-binding domain called the RFX motif. Multiple alternatively spliced transcript variants have been found but the full-length natures of only two have been determined. [provided by RefSeq]
Sequence: MAEDEPDAKSPKTGGRAPPGGAEAGEPTTLLQRLRGTISKAVQNKVEGILQDVQKFSDNDKLYLYLQLPSGPTTGDKSSEPSTLSNEEYMYAYRWIRNHLEEHTDTCLPKQSVYDAYRKYCESLACCRPLSTANFGKIIREIFPDIKARRLGGRGQSKYCYSGIRRKTLVSMPPLPGLDLKGSESPEMGPEVTPAPRDELVEAACALTCDWAERILKRSFSSIVEVARFLLQQHLISARSAHAHVLKAMGLAEEDEHAPRERSSKPKNGLENPEGGAHKKPERLAQPPKDLEARTGAGPLARGERKKSVVESSAPGANNLQVNALVARLPLLLPRAPRSLIPPIPVSPPILAPRLSSGALKVATLPLSSRAGAPPAAVPIINMILPTVPALPGPGPGPGRAPPGGLTQPRGTENREVGIGGDQGPHDKGVKRTAEVPVSEASGQAPPAKAAKQDIEDTASDAKRKRGRPRKKSGGSGERNSTPLKSAAAMESAQSSRLPWETWGSGGEGNSAGGAERPGPMGEAEKGAVLAQGQGDGTVSKGGRGPGSQHTKEAEDKIPLVPSKVSVIKGSRSQKEAFPLAKGEVDTAPQGNKDLKEHVLQSSLSQEHKDPKATPPSpecifications
NP_000440.1 | |
Antibody Production, Array, Enzyme-linked Immunoabsorbent Assay, Western Blot (Recombinant protein) | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
91.7kDa | |
Glutathione Sepharose 4 Fast Flow | |
25 ug | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
RFX5 | |
Wheat Germ (in vitro) | |
GST |
wheat germ expression system | |
Liquid | |
5993 | |
RFX5 (Human) Recombinant Protein (P01) | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
MAEDEPDAKSPKTGGRAPPGGAEAGEPTTLLQRLRGTISKAVQNKVEGILQDVQKFSDNDKLYLYLQLPSGPTTGDKSSEPSTLSNEEYMYAYRWIRNHLEEHTDTCLPKQSVYDAYRKYCESLACCRPLSTANFGKIIREIFPDIKARRLGGRGQSKYCYSGIRRKTLVSMPPLPGLDLKGSESPEMGPEVTPAPRDELVEAACALTCDWAERILKRSFSSIVEVARFLLQQHLISARSAHAHVLKAMGLAEEDEHAPRERSSKPKNGLENPEGGAHKKPERLAQPPKDLEARTGAGPLARGERKKSVVESSAPGANNLQVNALVARLPLLLPRAPRSLIPPIPVSPPILAPRLSSGALKVATLPLSSRAGAPPAAVPIINMILPTVPALPGPGPGPGRAPPGGLTQPRGTENREVGIGGDQGPHDKGVKRTAEVPVSEASGQAPPAKAAKQDIEDTASDAKRKRGRPRKKSGGSGERNSTPLKSAAAMESAQSSRLPWETWGSGGEGNSAGGAERPGPMGEAEKGAVLAQGQGDGTVSKGGRGPGSQHTKEAEDKIPLVPSKVSVIKGSRSQKEAFPLAKGEVDTAPQGNKDLKEHVLQSSLSQEHKDPKATPP | |
RUO | |
RFX5 | |
Yes |
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.