Learn More
Abnova™ Human RHBDL2 Partial ORF (NP_060291.2, 1 a.a. - 72 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
Supplier: Abnova™ H00054933Q01L
Description
The protein encoded by this gene is a member of the rhomboid family of integral membrane proteins. This family contains proteins that are related to Drosophila rhomboid protein. Members of this family are found in both prokaryotes and eukaryotes and are thought to function as intramembrane serine proteases. The encoded protein is thought to release soluble growth factors by proteolytic cleavage of certain membrane-bound substrates, including ephrin B2 and ephrin B3. [provided by RefSeq]
Sequence: MAAVHDLEMESMNLNMGREMKEELEEEEKMREDGGGKDRAKSKKVHRIVSKWMLPEKSRGTYLERANCFPPPSpecifications
NP_060291.2 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
33.66kDa | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
MAAVHDLEMESMNLNMGREMKEELEEEEKMREDGGGKDRAKSKKVHRIVSKWMLPEKSRGTYLERANCFPPP | |
RUO | |
RHBDL2 | |
Wheat Germ (in vitro) | |
GST | |
Liquid |
Antibody Production, ELISA, Protein Array, Western Blot | |
54933 | |
RHBDL2 (Human) Recombinant Protein (Q01) | |
25 ug | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
MGC16997/RRP2 | |
RHBDL2 | |
Recombinant | |
wheat germ expression system |
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.