Learn More
Novus Biologicals™ Human RhoGDI ELISA Kit (Colorimetric)
SDP
ELISA kits are ideal for the quantification of target antigens in tissue extracts, serum and cell culture.
Supplier:  Novus Biologicals™ NBP269952
Catalog No. NBP269952
Add to Cart
Description
Description
The Human RhoGDI ELISA Kit (Colorimetric) measures human RhoGDI in serum, plasma and other biological fluids.
Specifications
Specifications
0.16 - 10 ng/mL
This kit recognizes Human ARHGDI(alpha) in samples. No significant cross-reactivity or interference between Human ARHGDI(alpha) and analogues was observed.
Biological Fluids, Plasma, Serum
396
Human
Storage of components varies. See protocol for specific instructions.
0.10 ng/mL
ELISA Kit (Colorimetric)
ELISA
ARHGDIA
1 Kit
RhoGDI
Recommendations
Recommendations