Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Abnova Human RHOT1 Partial ORF (NP_060777, 483 a.a. - 580 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
Manufacturer: Abnova H00055288Q01S
Description
Sequence: TEAEIICDVVCLVYDVSNPKSFEYCARIFKQHFMDSRIPCLIVAAKSDLHEVKQEYSISPTDFCRKHKMPPPQAFTCNTADAPSKDIFVKLTTMAMYPSpecifications
NP_060777 | |
Antibody Production, Enzyme-linked Immunoabsorbent Assay, Protein Array, Western Blot (Recombinant protein) | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
36.52kDa | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
TEAEIICDVVCLVYDVSNPKSFEYCARIFKQHFMDSRIPCLIVAAKSDLHEVKQEYSISPTDFCRKHKMPPPQAFTCNTADAPSKDIFVKLTTMAMYP | |
RUO | |
RHOT1 | |
Wheat Germ (in vitro) | |
GST |
wheat germ expression system | |
Liquid | |
55288 | |
RHOT1 (Human) Recombinant Protein (Q01) | |
10 ug | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
ARHT1|FLJ11040|FLJ12633|MIRO-1 | |
RHOT1 | |
Yes |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title