Learn More
Abnova™ Human RNF121 Partial ORF (NP_060790.2, 193 a.a. - 301 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
$499.00 - $769.00
Specifications
Accession Number | NP_060790.2 |
---|---|
For Use With (Application) | Antibody Production, ELISA, Protein Array, Western Blot |
Formulation | 50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. |
Gene ID (Entrez) | 55298 |
Molecular Weight (g/mol) | 37.73kDa |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
89-960-169
|
Abnova™
H00055298Q01L |
25 ug |
Each for $769.00
|
|
|||||
89-960-168
|
Abnova™
H00055298Q01S |
10 ug |
Each for $499.00
|
|
|||||
Description
The protein encoded by this gene contains a RING finger, a motif present in a variety of functionally distinct proteins and known to be involved in protein-protein and protein-DNA interactions. Several alternatively spliced transcript variants have been noted for this gene, however, not all are likely to encode viable protein products. [provided by RefSeq]
Sequence: ERDFAEMCADYMASTIGFYSESGMPTKHLSDSVCAVCGQQIFVDVSEEGIIENTYRLSCNHVFHEFCIRGWCIVGKKQTCPYCKEKVDLKRMFSNPWERPHVMYGQLLDSpecifications
NP_060790.2 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
37.73kDa | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
FLJ11099 | |
RNF121 | |
Recombinant | |
wheat germ expression system |
Antibody Production, ELISA, Protein Array, Western Blot | |
55298 | |
RNF121 (Human) Recombinant Protein (Q01) | |
ERDFAEMCADYMASTIGFYSESGMPTKHLSDSVCAVCGQQIFVDVSEEGIIENTYRLSCNHVFHEFCIRGWCIVGKKQTCPYCKEKVDLKRMFSNPWERPHVMYGQLLD | |
RUO | |
RNF121 | |
Wheat Germ (in vitro) | |
GST | |
Liquid |
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.