Learn More
BioworldSupplier Diversity Partner HUMAN RO-60/SS-A (RECOMBINANT) DFS Item
Manufacturer:  BioworldSupplier Diversity Partner 220604721
Recombinant Human RO-60/SS-A Source: E.Coli Background: The SS-A/Ro particle contains a hY RNA (human cytoplasmic RNA) and associated proteins: a 60 kDa and a 52 kDa protein. The 52 kDa protein is not directly bound to the hY RNA but...
Catalog No. 50-253-568