Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abnova™ Human RP5-1022P6.2 Full-length ORF (NP_062539.1, 1 a.a. - 672 a.a.) Recombinant Protein with GST-tag at N-terminal

Used for AP, Array, ELISA, WB-Re

Manufacturer:  Abnova™ H00056261P01S

 View more versions of this product

Catalog No. 89-960-446


Add to cart

Description

Description

Sequence: MTPSQVAFEIRGTLLPGEVFAICGSCDALGNWNPQNAVALLPENDTGESMLWKATIVLSRGVSVQYRYFKGYFLEPKTIGGPCQVIVHKWETHLQPRSITPLESEIIIDDGQFGIHNGVETLDSGWLTCQTEIRLRLHYSEKPPVSITKKKLKKSRFRVKLTLEGLEEDDDDRVSPTVLHKMSNSLEISLISDNEFKCRHSQPECGYGLQPDRWTEYSIQTMEPDNLELIFDFFEEDLSEHVVQGDALPGHVGTACLLSSTIAESGKSAGILTLPIMSRNSRKTIGKVRVDYIIIKPLPGYSCDMKSSFSKYWKPRIPLDVGHRGAGNSTTTAQLAKVQENTIASLRNAASHGAAFVEFDVHLSKDFVPVVYHDLTCCLTMKKKFDADPVELFEIPVKELTFDQLQLLKLTHVTALKSKDRKESVVQEENSFSENQPFPSLKMVLESLPEDVGFNIEIKWICQQRDGMWDGNLSTYFDMNLFLDIILKTVLENSGKRRIVFSSFDADICTMVRQKQNKYPILFLTQGKSEIYPELMDLRSRTTPIAMSFAQFENLLGINVHTEDLLRNPSYIQEAKAKGLVIFCWGDDTNDPENRRKLKELGVNGLIYDRIYDWMPEQPNIFQVEQLERLKQELPELKSCLCPTVSRFVPSSLCGESDIHVDANGIDNVENA
Specifications

Specifications

wheat germ expression system
Antibody Production, Array, Enzyme-linked Immunoabsorbent Assay, Western Blot (Recombinant protein)
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
102.4kDa
Glutathione Sepharose 4 Fast Flow
10 ug
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
FLJ11085/KIAA1434/MGC26147/PREI4
RP5-1022P6.2
Yes
NP_062539.1
Liquid
56261
RP5-1022P6.2 (Human) Recombinant Protein (P01)
12.5% SDS-PAGE Stained with Coomassie Blue.
MTPSQVAFEIRGTLLPGEVFAICGSCDALGNWNPQNAVALLPENDTGESMLWKATIVLSRGVSVQYRYFKGYFLEPKTIGGPCQVIVHKWETHLQPRSITPLESEIIIDDGQFGIHNGVETLDSGWLTCQTEIRLRLHYSEKPPVSITKKKLKKSRFRVKLTLEGLEEDDDDRVSPTVLHKMSNSLEISLISDNEFKCRHSQPECGYGLQPDRWTEYSIQTMEPDNLELIFDFFEEDLSEHVVQGDALPGHVGTACLLSSTIAESGKSAGILTLPIMSRNSRKTIGKVRVDYIIIKPLPGYSCDMKSSFSKYWKPRIPLDVGHRGAGNSTTTAQLAKVQENTIASLRNAASHGAAFVEFDVHLSKDFVPVVYHDLTCCLTMKKKFDADPVELFEIPVKELTFDQLQLLKLTHVTALKSKDRKESVVQEENSFSENQPFPSLKMVLESLPEDVGFNIEIKWICQQRDGMWDGNLSTYFDMNLFLDIILKTVLENSGKRRIVFSSFDADICTMVRQKQNKYPILFLTQGKSEIYPELMDLRSRTTPIAMSFAQFENLLGINVHTEDLLRNPSYIQEAKAKGLVIFCWGDDTNDPENRRKLKELGVNGLIYDRIYDWMPEQPNIFQVEQLERLKQELPELKSCLCPTVSRFVPSSLCGESDIHVDANGIDNVENA
RUO
RP5-1022P6.2
Wheat Germ (in vitro)
GST
SDS
Documents
Product Certifications
Promotions

Promotions

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit