Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Human S100B His-tag, Recombinant Protein, Invitrogen™ Non-distribution product as customer accommodation.

E. coli Recombinant Protein

$1199.00

Specifications

Recombinant Yes
Name SARS-CoV-2 Nucleocapsid (aa1-419), His Tag
For Use With (Application) Control, ELISA, Western Blot
Conjugate Unlabeled
Format Lyophilized
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
PIRP87689 Non-distribution product as customer accommodation.
View Documents
Thermo Scientific
RP87689
200 µg
Each for $1,199.00
Add to cart
 
Description

Description

S100B is a member of the S100 family of proteins containing 2 EF-hand calcium-binding motifs. S100 proteins are localized in the cytoplasm and/or nucleus of a wide range of cells and are involved in the regulation of a number of cellular processes such as cell cycle progression and differentiation. S100B may function in neurite extension, proliferation of melanoma cells, stimulation of Ca2+ fluxes, inhibition of PKC-mediated phosphorylation, astrocytosis and axonal proliferation, and inhibition of microtubule assembly. Chromosomal rearrangements and altered expression of this gene have been implicated in several diseases, including Alzheimer's disease, Down's syndrome, epilepsy, amyotrophic lateral sclerosis, melanoma, and type I diabetes.
Specifications

Specifications

Yes
Control, ELISA, Western Blot
Lyophilized
Coronavirus; COVID; COVID 19; COVID19; COVID-19; SARS; SARS Coronavirus; SARS CoV; SARS CoV2; SARS CoV-2; SARS Nucleocapsid; SARS-CoV2; SARS-CoV-2
RUO
Virus
SARS-CoV-2 Nucleocapsid (aa1-419), His Tag
Unlabeled
PBS with no preservative
HEK293 cells
-20°Cor to +80°C if preferred
SDS
Product Certifications
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit