Learn More
Abnova™ Human SAC Partial ORF (NP_060887, 1504 a.a. - 1610 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
Supplier: Abnova™ H00055811Q01S
Description
The protein encoded by this gene belongs to a distinct class of mammalian adenylyl cyclase that is soluble and insensitive to G protein or forskolin regulation. It is localized in the cytoplasm and is thought to function as a general bicarbonate sensor throughout the body. It may also play an important role in the generation of cAMP in spermatozoa, implying possible roles in sperm maturation through the epididymis, capacitation, hypermotility, and/or the acrosome reaction. [provided by RefSeq]
Sequence: TTGPVFCPRLYHLMAYVCILMGDGQKCGLFLNTALRLSETQGNILEKCWLNMNKESWYSTSELKEDQWLQTILSLPSWEKIVAGRVNIQDLQKNKFLMRANTVDNHFSpecifications
NP_060887 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
37.51kDa | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
TTGPVFCPRLYHLMAYVCILMGDGQKCGLFLNTALRLSETQGNILEKCWLNMNKESWYSTSELKEDQWLQTILSLPSWEKIVAGRVNIQDLQKNKFLMRANTVDNHF | |
RUO | |
ADCY10 | |
Wheat Germ (in vitro) | |
GST | |
Liquid |
Antibody Production, ELISA, Protein Array, Western Blot | |
55811 | |
SAC (Human) Recombinant Protein (Q01) | |
10 ug | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
HCA2/RP1-313L4.2/SAC/SACI/Sacy | |
ADCY10 | |
Recombinant | |
wheat germ expression system |
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.