Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Abnova™ Human SAGE1 Partial ORF (NP_061136, 357 a.a. - 455 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
Supplier: Abnova™ H00055511Q01L
Description
This gene belongs to a class of genes that are activated in tumors. These genes are expressed in tumors of different histologic types but not in normal tissues, except for spermatogenic cells and, for some, placenta. The proteins encoded by these genes appear to be strictly tumor specific, and hence may be excellent sources of antigens for cancer immunotherapy. This gene is expressed in sarcomas. [provided by RefSeq]
Sequence: PLPSNALSTVLPGLAYLATADMPAMSTRDQHATIIHNLREEKKDNSQPTPDNVLSAVTPELINLAGAGIPPMSTRDQYATVNHHVHEARMENGQRKQDNSpecifications
NP_061136 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
36.63kDa | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
PLPSNALSTVLPGLAYLATADMPAMSTRDQHATIIHNLREEKKDNSQPTPDNVLSAVTPELINLAGAGIPPMSTRDQYATVNHHVHEARMENGQRKQDN | |
RUO | |
SAGE1 | |
Wheat Germ (in vitro) | |
GST | |
Liquid |
Antibody Production, ELISA, Protein Array, Western Blot | |
55511 | |
SAGE1 (Human) Recombinant Protein (Q01) | |
25 ug | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
SAGE | |
SAGE1 | |
Recombinant | |
wheat germ expression system |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction