Learn More
Invitrogen™ Human SARS-CoV-2 Spike (Trimer) IgM ELISA Kit
Manufacturer:  Invitrogen™ BMS2324TEN
Encompass
 View more versions of this product
Catalog No. BMS2324TEN
Add to cart
Description
Description
It has recently been shown that SARS (severe acute respiratory syndrome) is caused by a human coronavirus. Human coronaviruses are the major cause of upper respiratory tract illness, such as the common cold, in humans. Coronaviruses are positive-stranded RNA viruses, featuring the largest viral RNA genomes known to date (27-31 kb). The first step in coronavirus infection is binding of the viral spike protein, a 139-kDa protein, to certain receptors on host cells. The spike protein is the main surface antigen of the coronavirus. The glycosylated spike protein (as well as the nucleocapsid protein) can be detected in infected cell culture supernatants with antisera from SARS patients.
Specifications
Specifications
ELISA
Biotin
234 to 15,000 Units/mL
Research Use Only
2°C to 8°C
1 hr 15 min
<10%
10 x 96 Tests
<234 Units/mL
Colorimetric Microplate Reader
HRP
Pre-coated 96 well plate, High Control (standard), Calibrator (medium control), Low Control, IgM - HRP conjugate, Assay Buffer concentrate, Wash Buffer concentrate, Substrate solution, Stop Solution, Adhesive Plate Covers
Serum, 5 μL; Plasma, 5 μL
30 min
SARS-CoV-2 Spike (trimer) Protein
<10%
Serum, Plasma
Coronavirus; COVID; Covid 19; COVID19; COVID-19; SARS; SARS Coronavirus; SARS CoV; SARS CoV2; SARS CoV-2; SARS Spike Protein; SARS-CoV-2