Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abnova™ Human SARS2 Full-length ORF (NP_060297.1, 1 a.a. - 518 a.a.) Recombinant Protein with GST-tag at N-terminal

Used for AP, Array, ELISA, WB-Re

$460.50 - $709.50

Specifications

Name SARS2 (Human) Recombinant Protein (P01)
Recombinant Yes
Purification Method Glutathione Sepharose 4 Fast Flow
For Use With (Application) Antibody Production, Array, Enzyme-linked Immunoabsorbent Assay, Western Blot (Recombinant protein)
Molecular Weight 84.7kDa
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
89-961-073
View Documents
Abnova™
H00054938P01L
25 ug
Each for $709.50
Add to cart
 
89-961-072
View Documents
Abnova™
H00054938P01S
10 ug
Each for $460.50
Add to cart
 
Description

Description

This gene encodes the mitochondrial seryl-tRNA synthethase precursor, a member of the class II tRNA synthetase family. The mature enzyme catalyzes the ligation of Serine to tRNA(Ser) and participates in the biosynthesis of selenocysteinyl-tRNA(sec) in mitochondria. The enzyme contains an N-terminal tRNA binding domain and a core catalytic domain. It functions in a homodimeric form, which is stabilized by tRNA binding. This gene is regulated by a bidirectional promoter that also controls the expression of mitochondrial ribosomal protein S12. Both genes are within the critical interval for the autosomal dominant deafness locus DFNA4 and might be linked to this disease. Multiple transcript variants encoding different isoforms have been identified for this gene. [provided by RefSeq]

Sequence: MAASMARRLWPLLTRRGFRPRGGCISNDSPRRSFTTEKRNRNLLYEYAREGYSALPQLDIERFCACPEEAAHALELRKGELRSADLPAIISTWQELRQLQEQIRSLEEEKAAVTEAVRALLANQDSGEVQQDPKYQGLRARGREIRKELVHLYPREAQLEEQFYLQALKLPNQTHPDVPVGDESQARVLHMVGDKPVFSFQPRGHLEIGEKLDIIRQKRLSHVSGHRSYYLRGAGALLQHGLVNFTFNKLLRRGFTPMTVPDLLRGAVFEGCGMTPNANPSQIYNIDPARFKDLNLAGTAEVGLAGYFMDHTVAFRDLPVRMVCSSTCYRAETNTGQEPRGLYRVHHFTKVEMFGVTGPGLEQSSQLLEEFLSLQMEILTELGLHFRVLDMPTQELGLPAYRKFDIEAWMPGRGRFGEVTSASNCTDFQSRRLHIMFQTEAGELQFAHTVNATACAVPRLLIALLESNQQKDGSVLVPPALQSYLGTDRITAPTHVPLQYIGPNQPRKPGLPGQPAVS
Specifications

Specifications

SARS2 (Human) Recombinant Protein (P01)
Glutathione Sepharose 4 Fast Flow
84.7kDa
Yes
Antibody Production, Array, Enzyme-linked Immunoabsorbent Assay, Western Blot (Recombinant protein)
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
SDS
Product Certifications
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit