Learn More
Abnova™ Human SCAND2 Partial ORF (NP_071333, 45 a.a. - 134 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
Supplier: Abnova™ H00054581Q01S
Description
The SCAN domain is a highly conserved, leucine-rich motif of approximately 60 aa originally found within a subfamily of zinc finger proteins. This gene belongs to a family of genes that encode an isolated SCAN domain, but no zinc finger motif. Functional studies have established that the SCAN box is a protein interaction domain that mediates both hetero- and homoprotein associations, and maybe involved in regulation of transcriptional activity. Multiple transcript variants which encode the same isoform but differ only in their 3' UTRs, and another variant which encodes a distinct isoform have been described for this gene.
Sequence: CQSFWHFRYQEASRPREALLQLQKLCCQWLRPEKCTKEQILELLVLEQFPTVLLQEIQIWVRQQHPESGEEAVALVEDLQKEPGRQRLEPSpecifications
NP_071333 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
35.64kDa | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
CQSFWHFRYQEASRPREALLQLQKLCCQWLRPEKCTKEQILELLVLEQFPTVLLQEIQIWVRQQHPESGEEAVALVEDLQKEPGRQRLEP | |
RUO | |
SCAND2 | |
Recombinant | |
wheat germ expression system |
Antibody Production, ELISA, Protein Array, Western Blot | |
54581 | |
SCAND2 (Human) Recombinant Protein (Q01) | |
10 ug | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
SCAND2 | |
Wheat Germ (in vitro) | |
GST | |
Liquid |
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.