Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ CD23 (Soluble) Human ELISA Kit Non-distribution product as customer accommodation.

For the quantitative detection of human sCD23.

Manufacturer:  Invitrogen™ BMS2272

 View more versions of this product

Catalog No. 50-180-83


Add to cart

Includes: Aluminium pouch(es) with a Microwell Plate coated with monoclonal antibody to human sCD23
Biotin-Conjugate anti-human sCD23 monoclonal antibody
Streptavidin-HRP
Human sCD23 Standard lyophilized, 400 U/ml upon reconstitution
Control high, lyophilized
Control low, lyophilized
Assay Buffer Concentrate 20x (PBS with 1% Tween 20 and 10% BSA)
Wash Buffer Concentrate 20x (PBS with 1% Tween 20)
Substrate Solution (tetramethyl-benzidine)
Stop Solution (1M Phosphoric acid)
Blue-Dye
Green-Dye
Red-Dye
Adhesive Films

Description

Description

CD23 is a 45 kDa glycoprotein which is present on a subpopulation of freshly isolated peripheral blood and tonsil B cells and strongly expressed on EBV-transformed B lymphoblasts. The CD23 molecule is identical to the low affinity IgE receptor found on B cells. Expression of CD23 has been detected in neoplastic cells from cases of B cell chronic lymphocyctic leukaemia and some cases of centroblastic/centrocytic lymphoma. CD23 is present on a subpopulation of freshly isolated peripheral blood and tonsil B cells and strongly expressed on EBV-transformed B lymphoblasts. Functionally, CD23 is involved in B cell growth and differentiation, and IgE production. Further, CD23 has a soluble form that is a potent mitogenic factor. Diseases associated with CD23 dysfunction include chronic conjunctivitis and chronic lymphocytic leukemia.
Specifications

Specifications

P06734
Biotin
2208
ELISA Kit
6.8 U/mL
Plasma, 100 μL|Serum, 100 μL|Supernatant, 100 μL
1 hr. 20 min.
Human
Immunoglobulin E receptor low affinity II
4%
Human
HRP
12.5-400 U/mL
ELISA
Adhesive Plate Covers
RUO
100 μL
2°C to 8°C
3 hr. 10 min.
BLAST-2,CD23,CD23A,CLEC4J,FCE2,IGEBF
6.3%
96 Tests
Plasma, Serum, Supernatant
Colorimetric Microplate Reader
SDS
Documents
Provide Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit

For Research Use Only