Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Abnova™ Human SEC31B Full-length ORF (AAH44569.1, 1 a.a. - 348 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
Supplier: Abnova™ H00025956P01L
Description
This gene encodes a protein of unknown function. The protein has moderate similarity to rat VAP1 protein which is an endosomal membrane-associated protein, containing a putative Ca2+/calmodulin-dependent kinase II phosphorylation site. [provided by RefSeq]
Sequence: MKLKELERPAVQAWSPASQYPLYLATGTSAQQLDSSFSTNGTLEIFEVDFRDPSLDLKHRGVLSALSRFHKLVWGSFGSGLLESSGVIAGGGDNGMLILYNVTHILSSGKEPVIAQKQKHTGAVRALDFNPFQGNLLASGASDSEIFIWDLNNLNVPMTLGSKSQPPEDIKALSWNRQAQHILSSAHPSGKAVVWDLRKNEPIIKVSDHSNRMHCSGLAWHPDIATQLVLCSEDDRLPVIQLWDLRFASSPLKVLESHSRGILSVSWSQADAELLLTSAKDSQILCRNLGSSEVVYKLPTQSSWCFDVQWCPRDPSVFSAASFNGWISLYAVMGRSWEVQHMRQADKVSpecifications
AAH44569.1 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
64.8kDa | |
Glutathione Sepharose 4 Fast Flow | |
25 ug | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
DKFZp434M183/SEC31B-1/SEC31L2 | |
SEC31B | |
Recombinant | |
wheat germ expression system |
Antibody Production, Protein Array, ELISA, Western Blot | |
25956 | |
SEC31B (Human) Recombinant Protein (P01) | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
MKLKELERPAVQAWSPASQYPLYLATGTSAQQLDSSFSTNGTLEIFEVDFRDPSLDLKHRGVLSALSRFHKLVWGSFGSGLLESSGVIAGGGDNGMLILYNVTHILSSGKEPVIAQKQKHTGAVRALDFNPFQGNLLASGASDSEIFIWDLNNLNVPMTLGSKSQPPEDIKALSWNRQAQHILSSAHPSGKAVVWDLRKNEPIIKVSDHSNRMHCSGLAWHPDIATQLVLCSEDDRLPVIQLWDLRFASSPLKVLESHSRGILSVSWSQADAELLLTSAKDSQILCRNLGSSEVVYKLPTQSSWCFDVQWCPRDPSVFSAASFNGWISLYAVMGRSWEVQHMRQADKV | |
RUO | |
SEC31B | |
Wheat Germ (in vitro) | |
GST | |
Liquid |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction