Learn More
Abnova™ Human SEMA3A Partial ORF (NP_006071, 672 a.a. - 770 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
Supplier: Abnova™ H00010371Q01L
Description
This gene is a member of the semaphorin family and encodes a protein with an Ig-like C2-type (immunoglobulin-like) domain, a PSI domain and a Sema domain. This secreted protein can function as either a chemorepulsive agent, inhibiting axonal outgrowth, or as a chemoattractive agent, stimulating the growth of apical dendrites. In both cases, the protein is vital for normal neuronal pattern development. Increased expression of this protein is associated with schizophrenia and is seen in a variety of human tumor cell lines. Also, aberrant release of this protein is associated with the progression of Alzheimer's disease. [provided by RefSeq]
Sequence: HLEELLHKDDDGDGSKTKEMSNSMTPSQKVWYRDFMQLINHPNLNTMDEFCEQVWKRDRKQRRQRPGHTPGNSNKWKHLQENKKGRNRRTHEFERAPRSSpecifications
NP_006071 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
36.63kDa | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
HLEELLHKDDDGDGSKTKEMSNSMTPSQKVWYRDFMQLINHPNLNTMDEFCEQVWKRDRKQRRQRPGHTPGNSNKWKHLQENKKGRNRRTHEFERAPRS | |
RUO | |
SEMA3A | |
Wheat Germ (in vitro) | |
GST | |
Liquid |
Antibody Production, ELISA, Protein Array, Western Blot | |
10371 | |
SEMA3A (Human) Recombinant Protein (Q01) | |
25 ug | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
Hsema-I/Hsema-III/MGC133243/SEMA1/SEMAD/SEMAIII/SEMAL/SemD/coll-1 | |
SEMA3A | |
Recombinant | |
wheat germ expression system |
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.