Learn More
Abnova™ Human SENP7 Partial ORF (NP_065705, 1 a.a. - 110 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
Supplier: Abnova™ H00057337Q01S
Description
The reversible posttranslational modification of proteins by the addition of small ubiquitin-like SUMO proteins (see SUMO1; MIM 601912) is required for many cellular processes. SUMO-specific proteases, such as SENP7, process SUMO precursors to generate a C-terminal diglycine motif required for the conjugation reaction. They also display isopeptidase activity for deconjugation of SUMO-conjugated substrates (Lima and Reverter, 2008 [PubMed 18799455]).[supplied by OMIM]
Sequence: MDKRKLGRRPSSSEIITEGKRKKSSSDLSEIRKMLNAKPEDVHVQSPLSKFRSSERWTLPLQWERSLRNKVISLDHKNKKHIRGCPVTSRSSPERIPRVILTNVLGTELGSpecifications
NP_065705 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
37.84kDa | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
MDKRKLGRRPSSSEIITEGKRKKSSSDLSEIRKMLNAKPEDVHVQSPLSKFRSSERWTLPLQWERSLRNKVISLDHKNKKHIRGCPVTSRSSPERIPRVILTNVLGTELG | |
RUO | |
SENP7 | |
Wheat Germ (in vitro) | |
GST | |
Liquid |
Antibody Production, ELISA, Protein Array, Western Blot | |
57337 | |
SENP7 (Human) Recombinant Protein (Q01) | |
10 ug | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
KIAA1707/MGC157730 | |
SENP7 | |
Recombinant | |
wheat germ expression system |
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.