Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abnova™ Human SENP7 Partial ORF (NP_065705, 1 a.a. - 110 a.a.) Recombinant Protein with GST-tag at N-terminal

Used for AP, Array, ELISA, WB-Re

Manufacturer:  Abnova™ H00057337Q01L

 View more versions of this product

Catalog No. 89-961-441


Add to cart

Description

Description

The reversible posttranslational modification of proteins by the addition of small ubiquitin-like SUMO proteins (see SUMO1; MIM 601912) is required for many cellular processes. SUMO-specific proteases, such as SENP7, process SUMO precursors to generate a C-terminal diglycine motif required for the conjugation reaction. They also display isopeptidase activity for deconjugation of SUMO-conjugated substrates (Lima and Reverter, 2008 [PubMed 18799455]).[supplied by OMIM]

Sequence: MDKRKLGRRPSSSEIITEGKRKKSSSDLSEIRKMLNAKPEDVHVQSPLSKFRSSERWTLPLQWERSLRNKVISLDHKNKKHIRGCPVTSRSSPERIPRVILTNVLGTELG
Specifications

Specifications

wheat germ expression system
Antibody Production, Enzyme-linked Immunoabsorbent Assay, Protein Array, Western Blot (Recombinant protein)
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
37.84kDa
12.5% SDS-PAGE Stained with Coomassie Blue.
MDKRKLGRRPSSSEIITEGKRKKSSSDLSEIRKMLNAKPEDVHVQSPLSKFRSSERWTLPLQWERSLRNKVISLDHKNKKHIRGCPVTSRSSPERIPRVILTNVLGTELG
RUO
SENP7
Wheat Germ (in vitro)
GST
NP_065705
Liquid
57337
SENP7 (Human) Recombinant Protein (Q01)
25 ug
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
KIAA1707/MGC157730
SENP7
Yes
SDS
Documents
Product Certifications
Promotions

Promotions

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit