Learn More
Abnova™ Human SEPHS2 Partial ORF (NP_036380.2, 61 a.a. - 150 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
$499.00 - $769.00
Specifications
Accession Number | NP_036380.2 |
---|---|
For Use With (Application) | Antibody Production, ELISA, Protein Array, Western Blot |
Formulation | 50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. |
Gene ID (Entrez) | 22928 |
Molecular Weight (g/mol) | 35.53kDa |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
89-961-453
|
Abnova™
H00022928Q01L |
25 ug |
Each for $769.00
|
|
|||||
89-961-452
|
Abnova™
H00022928Q01S |
10 ug |
Each for $499.00
|
|
|||||
Description
This gene encodes an enzyme that synthesizes selenophosphate from selenide and ATP. Selenophosphate is the selenium donor used to synthesize selenocysteine, which is co-translationally incorporated into selenoproteins at in-frame UGA codons. This protein itself contains a selenocysteine residue in its predicted active site. The 3' UTR of the gene has a stem-loop secondary structure called a selenocysteine insertion sequence (SECIS) element, which allows UGA to direct the incorporation of selenocysteine rather than signal a translational stop. Alternatively spliced transcripts have been identified, but their biological validity has not been determined. [provided by RefSeq]
Sequence: GCKVPQEALLKLLAGLTRPDVRPPLGRGLVGGQEEASQEAGLPAGAGPSPTFPALGIGMDSCVIPLRHGGLSLVQTTDFFYPLVEDPYMMSpecifications
NP_036380.2 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
35.53kDa | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
SPS2 | |
SEPHS2 | |
Recombinant | |
wheat germ expression system |
Antibody Production, ELISA, Protein Array, Western Blot | |
22928 | |
SEPHS2 (Human) Recombinant Protein (Q01) | |
GCKVPQEALLKLLAGLTRPDVRPPLGRGLVGGQEEASQEAGLPAGAGPSPTFPALGIGMDSCVIPLRHGGLSLVQTTDFFYPLVEDPYMM | |
RUO | |
SEPHS2 | |
Wheat Germ (in vitro) | |
GST | |
Liquid |
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.